Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Stanbio™ RELY™ Mono Rapid Test Kit
For qualitative detection of infectious mononucleosis
Includes: Individually pouched cassettes, sample buffer, disposable sample droppers, disposable heparinized capillary tubes with dispensing bulb and positive and negative controls, and easy-to-use procedure card
  • Two step test
  • Provides results in 5 minutes
  • Suitable for use with multiple sample types
View Specifications
Images for Stanbio™ RELY™ Mono Rapid Test Kit
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
22-022-518
STANBIO LABORATORY INC
No.:6200-020
Mononucleosis Rapid Test Kit
Each for $219.05
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Two step test
  • Provides results in 5 minutes
  • Suitable for use with multiple sample types
Certifications: CLIA waived for whole blood sample
Note: For in-vitro diagnostic use only