Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
S06695
Products for Science Education Large Molecular Model Set
For middle school through high school chemistry courses in which students learn about inorganic and organic molecules, as well as crystal molecules like graphite and diamond.
Includes: Plastic storage case
  • The springs easily allow demonstrations of single bonds, double bonds and triple bonds
  • Ball and spoke molecular model set includes 176 plastic spheres, 225 rigid plastic connector tubes and 10 flexible tubes
  • Spheres are 30mm in diameter, except for hydrogen, which are 23mm in diameter
  • Holes on the molecules are properly placed to make realistic figures
View Specifications
Alert: Not for children under 3 years.
Images for Products for Science Education Large Molecular Model Set
  • S06695
S06695
S06695
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
S06695
Walter Products Inc.
No.:MOLE1004
Large molecular model set
Each for $79.00
Check Availability
Add
to Cart
(0) in My Cart
Description
  • The springs easily allow demonstrations of single bonds, double bonds and triple bonds
  • Ball and spoke molecular model set includes 176 plastic spheres, 225 rigid plastic connector tubes and 10 flexible tubes
  • Spheres are 30mm in diameter, except for hydrogen, which are 23mm in diameter
  • Holes on the molecules are properly placed to make realistic figures