Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
MP116004500
MP Biomedicals FastPrep -24™ Instrument
High-speed, digitally controlled benchtop homogenizer for the lysis of biological samples
Uses a unique, optimized motion to disrupt cells through multidirectional, simultaneous beating of specialized lysing matrix beads on the sample material.
  • Simultaneous homogenization of 24 samples within 40 seconds
  • Developed to lyse difficult and resistant samples
  • Lyses tissues and cells thoroughly and quickly allowing easy and reproducible isolation of stable RNA, active proteins and full length genomic DNA
  • Ergonomic design ensures easy loading of the sample tubes that remain securely sealed while processing
View Specifications
Images for MP Biomedicals FastPrep -24™ Instrument
  • MP116004500
MP116004500
MP116004500
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
MP116004500
MP Biomedicals
No.:116004500
Benchtop
N/A N/A N/A
Price File Not Open 
Description
  • Simultaneous homogenization of 24 samples within 40 seconds
  • Developed to lyse difficult and resistant samples
  • Lyses tissues and cells thoroughly and quickly allowing easy and reproducible isolation of stable RNA, active proteins and full length genomic DNA
  • Ergonomic design ensures easy loading of the sample tubes that remain securely sealed while processing