Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
anti-CD4-Biotin; Clone: 13B8.2; Beckman Coulter™
Host Species: Mouse
Clone: 13B8.2
Isotype: IgG1
Species Reactivity: Hu
Immunogen: Human thymocytes
View Specifications
Images for anti-CD4-Biotin; Clone: 13B8.2; Beckman Coulter™
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
COIM0704
Beckman Coulter
No.:IM0704
0.2mg; Biotin
Each for $394.00
Check Availability
Add
to Cart
(0) in My Cart
Description
Host Species: Mouse
Clone: 13B8.2
Isotype: IgG1
Species Reactivity: Hu
Immunogen: Human thymocytes
Applications: Flow Cytometry
Recommended Storage: 2 to 8°C