Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
07-202-150
Corning™ Mini Bioreactor Centrifuge Tube
Ideal for high throughput process optimization for suspension cell culture
Product consists of a 50mL centrifuge tube with a vented cap.
  • Polypropylene tube with large marking spot
  • Polypropylene cap with 4 vents (hydrophobic membrane provides gas exchange)
  • Sterile, nonpyrogenic and RNase-/DNase-free
View Specifications
Images for Corning™ Mini Bioreactor Centrifuge Tube
  • 07-202-150
07-202-150
07-202-150
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
07-202-150
Corning Life Sciences
No.:431720
50mL
Case of 300 for $417.00
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Polypropylene tube with large marking spot
  • Polypropylene cap with 4 vents (hydrophobic membrane provides gas exchange)
  • Sterile, nonpyrogenic and RNase-/DNase-free