Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
S68771
Products for Science Education Adjustable Tuning Fork
Precision-grade aluminum alloy.
  • Sliding adjustable weights on each prong
  • Marked to deliver tones at C-128, D-144, E-160, F-170.7, G-192, A-213.3 and B-240 frequencies
View Specifications
Images for Products for Science Education Adjustable Tuning Fork
  • S68771
S68771
S68771
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
S68771
Medivibe Technology
No.:700240ADJ
Adjustable Tuning Fork
Each for $14.75
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Sliding adjustable weights on each prong
  • Marked to deliver tones at C-128, D-144, E-160, F-170.7, G-192, A-213.3 and B-240 frequencies