Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Products for Science Education Chemistry Demonstration Kits: Acetylene-Chlorine Reaction
In this demonstration, calcium carbide is placed in a cylinder containing water. A reaction occurs producing acetylene gas bubbles. A plastic tube from a chlorine gas generator is placed in the cylinder. As the bubbles of acetylene and chlorine collide, flashes of light are produced. These flashes of light are actually explosions! Acetylene contains a high-energy C-C triple bond. When this bond is broken by the reaction with chlorine, a great deal of energy is rapidly released causing explosions.
View Specifications
Images for Products for Science Education Chemistry Demonstration Kits: Acetylene-Chlorine Reaction
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
S68309 Acetylene-Chlorine Reaction
Each for $32.50
Check Availability
Add
to Cart
(0) in My Cart
Description
In this demonstration, calcium carbide is placed in a cylinder containing water. A reaction occurs producing acetylene gas bubbles. A plastic tube from a chlorine gas generator is placed in the cylinder. As the bubbles of acetylene and chlorine collide, flashes of light are produced. These flashes of light are actually explosions! Acetylene contains a high-energy C-C triple bond. When this bond is broken by the reaction with chlorine, a great deal of energy is rapidly released causing explosions.