Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
15-060-237
Ertco™ PTFE Coated Glass Special-Application Red-Spirit™-Filled Thermometers
Individually serialized
Precision-grade thermometers in a variety of ranges and immersions
  • Individually serialized
  • Filled with custom organic red spirit instead of mercury
  • Glass with or without a PTFE coating
CERTIFICATIONS:
Each thermometer is individually calibrated against equipment whose calibration is traceable to NIST™.
View Specifications
Images for Ertco™ PTFE Coated Glass Special-Application Red-Spirit™-Filled Thermometers
  • 15-060-237
15-060-237
15-060-237
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
15-060-253
Thermo Scientific
No.:10970-05
Range: 35° to 200°C; 180mm L
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
15-060-243
Thermo Scientific
No.:10835-05
Range: 0° to 110°C; 225mm
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
15-060-237
Thermo Scientific
No.:10919-05
Range: 25deg./37deg./56deg.C; 240mm L
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
15-060-241
Thermo Scientific
No.:10832-05
Range: 25° to 57°C; 175mm L
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Add Items to Cart
Description
Precision-grade thermometers in a variety of ranges and immersions
  • Individually serialized
  • Filled with custom organic red spirit instead of mercury
  • Glass with or without a PTFE coating
CERTIFICATIONS:
Each thermometer is individually calibrated against equipment whose calibration is traceable to NIST™.
  • Filled with custom organic red spirit instead of mercury
  • Glass with or without a PTFE coating
Certifications: Traceability certificate is not included and must be purchased separately. Contact your Fisher Scientific Customer Service Representative for more information.
† Arrows on the scale indicate critical temperatures.