Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
View Image Gallery (2) Image(s) are representative of the product Group
  • acros
Amberjet* 4200-Cl, Acros Organics
Amberjet* 4200-Cl
[60177-39-1]
View Specifications
Images for Amberjet* 4200-Cl, Acros Organics
  • acros
(3 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
500g
MSDS
AC30133-5000
Acros Organics
No.:301335000
N/AYou must be logged in and have a valid account number to view item availability   Each for $69.30
(0) in My Cart
2.5kg
MSDS
AC30133-0025
Acros Organics
No.:301330025
N/AYou must be logged in and have a valid account number to view item availability   Each for $289.00
(0) in My Cart
100g
MSDS
AC30133-1000
Acros Organics
No.:301331000
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Add Items to Cart
Product Specifications
Physical FormYellow translucent beads
Particle Size0.600 to 0.800mm harmonic mean size, 0.5% max. (< 0.425mm), 5.0% max. (> 0.850mm), 1.25 uniformity coefficient max.
Moisture Holding Capacity(Cl): 49% to 55%
Volume Total Capacity(Cl): 1.30 eq/L min.