Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
15-184-42
Available on GSA/VA Contract for Federal Government customers only. TBS™ Slide Dryer II
The convenient way to dry slides
  • Temperature control to 75°C (167°F)
  • Accepts two slide racks directly from stainer
  • Blows warm air over slide surface to dry slides in 20 to 30 minutes
  • Self-resetting thermal switch protects unit from overheating
  • Stainless-steel hinged lid, metal housing and Lexan™ front for easy viewing
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. TBS™ Slide Dryer II
  • 15-184-42
15-184-42
15-184-42
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
15-184-42 Available on GSA/VA Contract for Federal Government customers only. 
Triangle Biomedical Sciences
No.:SD-II-120
Slide Dryer II
Each for $2,010.00
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Temperature control to 75°C (167°F)
  • Accepts two slide racks directly from stainer
  • Blows warm air over slide surface to dry slides in 20 to 30 minutes
  • Self-resetting thermal switch protects unit from overheating
  • Stainless-steel hinged lid, metal housing and Lexan™ front for easy viewing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.