Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
19-816-109
View Image Gallery (3) Image(s) are representative of the product Group
  • 19-816-109
  • Delta Hydro Fire Hose Tester
  • Delta Hydro Fire Hose Tester
Niedner™ Delta Hydro Fire Hose Tester
Unique check valve system closes automatically in the event of hose failure without shutting down the other lines
Fills hoses faster than an engine. Delivers up to 500psi of pressure. NPSH threads for use with Navy and Coast Guard 1.5 and 1.75 in. (38 and 45mm) hoses.
Includes: Inline filter and 1.5 in. (38mm) NPSH female swivel inlet connector included. Tester does not include pressure washing equipment.
  • All stainless-steel manifold
  • Low-profile, heavy-duty, stainless-steel cart
  • Powerful, 1.7hp, totally enclosed, fan-cooled industrial motor
  • Six outlets, each with a fully automatic check valve system
  • Large 3.2gpm (12.6L) oil bath pump, complete with thermal protective valve
View Specifications
Images for Niedner™ Delta Hydro Fire Hose Tester
  • 19-816-109
  • Delta Hydro Fire Hose Tester
  • Delta Hydro Fire Hose Tester
19-816-109
19-816-109
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
19-816-109
NIEDNER
No.:CDHT1500SS-BRA
1.5 in manifold; 1.7HP; 1.5 in Brass NPSH check valve; SS Cart
Each for $5,398.85
Check Availability
Add
to Cart
(0) in My Cart
19-823-825
NIEDNER
No.:HT2000SS
2 in. manifold; 1.7 HP
Each for $4,016.15
Check Availability
Add
to Cart
(0) in My Cart
19-130-5356
NIEDNER
No.:CDHT1500-BRA
1.5 in.manifold
Each for $5,966.84
Check Availability
Add
to Cart
(0) in My Cart
19-816-110
NIEDNER
No.:CDHT-2000-SS
2 in. SS manifold; 1.7HP; 2.5 in Aluminum NST check valves; SS Cart
Each for $9,129.87
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • All stainless-steel manifold
  • Low-profile, heavy-duty, stainless-steel cart
  • Powerful, 1.7hp, totally enclosed, fan-cooled industrial motor
  • Six outlets, each with a fully automatic check valve system
  • Large 3.2gpm (12.6L) oil bath pump, complete with thermal protective valve
Compliance: Meets NFPA standards.
Warranty: One-year limited warranty.