Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
(Top) 05-408-150, (Bottom) 05-408-151
Fisherbrand™ Premium Plus MultiFlex Gel-Loading Tips
Ideal for gel electrophoresis
  • Easily access bottom of narrow vessels and gel wells
  • Universal fit
  • Graduated marks for quick volume checks
  • Certified DNase-, RNase-, and human DNA-free
Match O.D. to Gel Thickness
  • Round 0.57mm O.D. tips are recommended for loading gels ≥0.6mm thick—great for SDS Page and 2-D gels
  • Flat tips are recommended for gels up to 0.4mm thick
  • Use flat 0.37mm O.D. tips for 0.4mm thick PAGE sequencing gels; flat 0.17mm tips for 0.2mm DNA sequencing gels
MultiFlex Tips
  • 0.5 to 200µL volume
  • Fit research-grade pipetters
  • 83mm long
  • 5µL graduation
View Specifications
Images for Fisherbrand™ Premium Plus MultiFlex Gel-Loading Tips
  • (Top) 05-408-150, (Bottom) 05-408-151
(Top) 05-408-150, (Bottom) 05-408-151
(Top) 05-408-150, (Bottom) 05-408-151
(3 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
05-408-150 Flat; 0.5 to 200µL; O.D.: 0.37mm; 400/cs.
Case of 400 for $143.54
Check Availability
Add
to Cart
(0) in My Cart
05-408-151 Round; 0.5 to 200µL; O.D.: 0.57mm; 400/Cs.
Case of 400 for $77.23
Check Availability
Add
to Cart
(0) in My Cart
05-408-154 Round; 0.5 to 200µL; O.D.: 0.57mm; 1000/bag
Pack of 1000 for $94.44
Case of 10 PK for $471.14
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Easily access bottom of narrow vessels and gel wells
  • Universal fit
  • Graduated marks for quick volume checks
  • Certified DNase-, RNase-, and human DNA-free
Match O.D. to Gel Thickness
  • Round 0.57mm O.D. tips are recommended for loading gels ≥0.6mm thick—great for SDS Page and 2-D gels
  • Flat tips are recommended for gels up to 0.4mm thick
  • Use flat 0.37mm O.D. tips for 0.4mm thick PAGE sequencing gels; flat 0.17mm tips for 0.2mm DNA sequencing gels
MultiFlex Tips
  • 0.5 to 200µL volume
  • Fit research-grade pipetters
  • 83mm long
  • 5µL graduation