Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
S90528
Products for Science Education Fisher Science Education™ pH Meters
Large, dual LCD shows pH measurement and temperature results simultaneously (with ATC probes).
  • pH Mode: 0.00 to 14.00pH with resolution of 0.01 and accuracy of ±0.01pH
  • Temperature Mode (with ATC probe): 0.0 to 100°C with resolution of 0.1°C and accuracy of 0.3°C
  • mV Mode (S90525, S90527 and S90528 only): -1000.0 to +1000.0 with a resolution of 0.1 and an accuracy of 0.1mV within ±199.9mV; 1mV beyond ±199.9mV
  • mV models also feature recorder output
  • Simple push-button standardization with auto-buffer recognition
  • Icons clearly show current meter status
  • Three-point calibration with percent slope calculation
  • Simple user-interface
View Specifications
Images for Products for Science Education Fisher Science Education™ pH Meters
  • S90528
S90528
S90528
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
S90524 Meter only, pH/temp. modes
Each for $472.18
Check Availability
Add
to Cart
(0) in My Cart
S90528 Includes electrode and swing arm
Each for $886.40
Check Availability
Add
to Cart
(0) in My Cart
S90527 Meter with electrode, pH/mV/temp. modes
Each for $607.73
Check Availability
Add
to Cart
(0) in My Cart
S90526 Meter with electrode, pH/temp. modes
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Add Items to Cart
Description
  • pH Mode: 0.00 to 14.00pH with resolution of 0.01 and accuracy of ±0.01pH
  • Temperature Mode (with ATC probe): 0.0 to 100°C with resolution of 0.1°C and accuracy of 0.3°C
  • mV Mode (S90525, S90527 and S90528 only): -1000.0 to +1000.0 with a resolution of 0.1 and an accuracy of 0.1mV within ±199.9mV; 1mV beyond ±199.9mV
  • mV models also feature recorder output
  • Simple push-button standardization with auto-buffer recognition
  • Icons clearly show current meter status
  • Three-point calibration with percent slope calculation
  • Simple user-interface