Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
15-077-963
Fisher Scientific™ Traceable™ Dew-Point/Wet-Bulb/Humidity Thermometer
Monitor shows current, minimum, and maximum displays
Includes: Two “AA” alkaline batteries
  • Temperature range, -40° to +70°C (-40° to +158°F), with ±1°C accuracy and 0.1° resolution (below 100)
  • Relative humidity range, 1 to 99%; accuracy, ±4% between 20 to 80% RH; resolution, 0.1% RH
  • Settable high/low alarm for all four readings (dew-point, wet-bulb, humidity, temperature) Alarm sounds and flashes; may be set in 0.1 increments Large LCD digits are 1 in. (2.5cm) high Case is high-impact, chemical-resistant ABS plastic
View Specifications
Images for Fisher Scientific™ Traceable™ Dew-Point/Wet-Bulb/Humidity Thermometer
  • 15-077-963
15-077-963
15-077-963
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
15-077-963 Dew-point/wet-bulb/humidity thermometer
Each for $69.00
Each for $65.55 Online Instant Savings (5%)
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Temperature range, -40° to +70°C (-40° to +158°F), with ±1°C accuracy and 0.1° resolution (below 100)
  • Relative humidity range, 1 to 99%; accuracy, ±4% between 20 to 80% RH; resolution, 0.1% RH
  • Settable high/low alarm for all four readings (dew-point, wet-bulb, humidity, temperature) Alarm sounds and flashes; may be set in 0.1 increments Large LCD digits are 1 in. (2.5cm) high Case is high-impact, chemical-resistant ABS plastic
Specifications & Ordering Information:
Dimensions: 4.12 x 4.12 x 1 in. (10.5 x 10.5 x 2.5cm); Weight: 4.25 oz.
Certifications: A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology (NIST™). Calibrated by an ISO™ 17025 calibration laboratory accredited by A2LA™.
Compliance: Meets federal requirements for CLIA specification 493.1207 for monitoring temperature and humidity.