Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
742 Series Cartridge
Scott Safety™ 742 Series Air-purifying Respirator Accessory, Cartridges
Easy-breathingtwin cartridges were developed to provide optimal respiratory protection in non-IDLH industrial or HAZMAT situations
  • Low profile, swept back cartridge design ensures optimal field of vision and allows use of communications
  • Available in a full range of NIOSH approved, chemical cartridges or combination cartridges
  • Attaches directly to XCEL Halfmask and Promask full facepiece
  • Uses Quarter turn adapter when used with AV Series Facepieces
View Specifications
Images for Scott Safety™ 742 Series Air-purifying Respirator Accessory, Cartridges
  • 742 Series Cartridge
742 Series Cartridge
742 Series Cartridge
(10 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
19-161-800
Scott Safety
No.:7422-BA1
Organic Vapor Cartridges
Pack for $12.00
Check Availability
Add
to Cart
(0) in My Cart
19-161-796
Scott Safety
No.:7422-FP1
P100 Filter
Pack for $10.00
Check Availability
Add
to Cart
(0) in My Cart
19-161-802
Scott Safety
No.:7422-YD1
Organic Vapor/Acid Gas/P100 Assemblies
Pack of 2 for $24.00
Check Availability
Add
to Cart
(0) in My Cart
19-150-1254
Scott Safety
No.:7422-SD1
Multi-Purpose/P100 Assemblies
Pack of 2 for $24.90
Check Availability
Add
to Cart
(0) in My Cart
19-161-801
Scott Safety
No.:7422-BB1
Organic Vapor/P100 Assemblies
Pack for $23.30
Check Availability
Add
to Cart
(0) in My Cart
19-161-797
Scott Safety
No.:7422-WA1
Acid Gas Cartridges
Pack for $12.90
Check Availability
Add
to Cart
(0) in My Cart
19-700-952
Scott Safety
No.:7422-PD1
TEDA (radioiodine), Chlorine/P100
Each for $37.70
Check Availability
Add
to Cart
(0) in My Cart
19-161-740
Scott Safety
No.:7422SC1
Multi-Purpose Cartridges
Pack of 2 for $13.10
Check Availability
Add
to Cart
(0) in My Cart
19-700-845
Scott Safety
No.:7422-WB1
Acid Gas/P100 Assemblies
Pack of 2 for $24.60
Check Availability
Add
to Cart
(0) in My Cart
19-161-798
Scott Safety
No.:7422-ZA1
Mercury Vapor/Chlorine Cartridges
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Add Items to Cart
Description
  • Low profile, swept back cartridge design ensures optimal field of vision and allows use of communications
  • Available in a full range of NIOSH approved, chemical cartridges or combination cartridges
  • Attaches directly to XCEL Halfmask and Promask full facepiece
  • Uses Quarter turn adapter when used with AV Series Facepieces