Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Available on GSA/VA Contract for Federal Government customers only. Fisher BioReagents™ Bovine Serum Albumin, Fraction V, Heat Shock Treated
Suitable for immunological studies
CAS: 9048-46-8
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Fisher BioReagents™ Bovine Serum Albumin, Fraction V, Heat Shock Treated
(2 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
BP1600-100 Available on GSA/VA Contract for Federal Government customers only.  100g
MSDS
Each for $240.68
Each for $192.54 Online Instant Savings (20%)
Case of 6 EA for $1,083.08
Case of 6 EA for $866.46 Online Instant Savings (20%)
Check Availability
Add
to Cart
(0) in My Cart
BP1600-1 Available on GSA/VA Contract for Federal Government customers only.  1kg
MSDS
Each for $1,463.94
Each for $1,171.15 Online Instant Savings (20%)
Case of 4 EA for $4,684.62
Case of 4 EA for $3,747.70 Online Instant Savings (20%)
Check Availability
Add
to Cart
(0) in My Cart
Description
CAS: 9048-46-8
Product Specifications
Assay>=98%
Heavy Metals (as Pb)<=0.002%
Loss on Drying at 105°C<=5%
pH (1% Solution in 0.9% NaCl)7.0 ±0.3
Solubility (0.4g/10mL H2O)Clear and haze-free
Sulfated Ash<=2%
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.