Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Lubrol MP Biomedicals
Lubrol
Molecular Biology
Off-white Waxy Solid
Polyethylene glycolmonocetyl ether
HO(CH2CH2O)n C16H33 n = ~23
[8047-15-2]
Images for Lubrol MP Biomedicals
  • mpbiomedical
(2 item(s) in product Group)
  • Items
  • Physical Properties
  • Safety and Handling
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
ICN19529980
MP Biomedicals
No.:0219529980
100g
Each for $95.30
Check Availability
Add
to Cart
(0) in My Cart
ICN19529983
MP Biomedicals
No.:0219529983
250g
Each for $131.60
Check Availability
Add
to Cart
(0) in My Cart
Melting Point: 38° to 48°C
Freely soluble in alcohols. Soluble in water.
Recommended storage: Store at room temperature (15° to 30°C)