Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Thermo Scientific Halt Phosphatase Inhibitor Cocktail
View Image Gallery (2) Image(s) are representative of the product Group
  • Thermo Scientific Halt Phosphatase Inhibitor Cocktail
  • Preserved phosphorylation states of three proteins in cell lysate.
Thermo Scientific™ Halt™ Phosphatase Inhibitor Cocktail
Safeguards against serine, threonine and protein tyrosine phosphatases activity
The Halt Phosphatase Inhibitor Cocktail protects phosphoproteins from serine/threonine phosphatases as well as protein tyrosine phosphatases (PTPs). The cocktail contains a mixture of four inhibitors of broad specificity, including sodium fluoride, sodium orthovanadate, sodium pyrophosphate, and β-glycerophosphate. Conveniently packaged in a 100X format, the cocktail is stable at 4°C and is suitable for use with any cell or tissue extract. The cocktail preserves protein phosphorylation in immunoprecipitation and kinase assays. Furthermore, the cocktail is compatible with popular protein assays such as the Thermo Scientific Pierce™ BCA Protein Assay Kit (Part No. 23225) and Bradford-based Pierce Coomassie Plus™ Assay Kit (Part No. 23236).
Highlights:
  • All-in-one format – inhibits both serine-threonine phosphatases and protein tyrosine phosphatases
  • Compatible – works with standard protein assays, including BCA and coomassie (Bradford) assays
  • Convenient storage – refrigerator (2 to 8°C) storage saves valuable space in the freezer and provides for immediate use
  • Multiple sizes – select a package size of the 100X cocktail that is ideal for any particular scale and workflow of protein extraction experiment
View Specifications
Images for Thermo Scientific™ Halt™ Phosphatase Inhibitor Cocktail
  • Thermo Scientific Halt Phosphatase Inhibitor Cocktail
  • Preserved phosphorylation states of three proteins in cell lysate.
Thermo Scientific Halt Phosphatase Inhibitor Cocktail
Thermo Scientific Halt Phosphatase Inhibitor Cocktail
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
PI-78420
Thermo Scientific
No.:78420
1mL
Each for $192.19
Check Availability
Add
to Cart
(0) in My Cart
PI-78426
Thermo Scientific
No.:78426
Cocktail; 5 x 1mL
Each for $585.81
Check Availability
Add
to Cart
(0) in My Cart
PI-78427
Thermo Scientific
No.:78427
Cocktail; 10mL
Each for $789.14
Check Availability
Add
to Cart
(0) in My Cart
PI-78428
Thermo Scientific
No.:78428
Cocktail; Single-use vials; 24 x 100µL
Each for $298.25
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
Highlights:
  • All-in-one format – inhibits both serine-threonine phosphatases and protein tyrosine phosphatases
  • Compatible – works with standard protein assays, including BCA and coomassie (Bradford) assays
  • Convenient storage – refrigerator (2 to 8°C) storage saves valuable space in the freezer and provides for immediate use
  • Multiple sizes – select a package size of the 100X cocktail that is ideal for any particular scale and workflow of protein extraction experiment