Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
S6307 Series
Fisher Scientific™ Isotemp™ Digital-Control Water Baths
Ideal for industrial, clinical, pharmaceutical or biomedical procedures
Ideal for industrial, clinical, pharmaceutical or biomedical applications
Microprocessor PID Control
  • ±0.24°C uniformity, ±0.5°C accuracy and stability at 37°C
  • Display shows °C or °F; easy to read from across the room
Safety Features
  • Low profile and front-panel controls for easier access, safer operation
  • Dual thermostats guard against thermal runaway
  • Hinged acrylic or stainless-steel gable covers have fins that keep hands away from hot vapors
  • Covers stay open at 90° or, with quick-release hinge, lift off completely for use with large glassware
  • Heating elements will not burn out if bath accidentally runs dry
Construction
  • Corrosion-resistant exterior with tough, easy-to-clean acrylic enamel coating
  • Temperature-resistant plastic rim insulates, resists leaks, keeps its shape and won't dry out or crack like caulked seals
  • Exterior stays cool even after hours of operation Deep-drawn, stainless-steel reservoirs and perforated sample trays prevent leaks, guard against rust and contamination
  • Rounded reservoir corners and sample trays with finger holes allow for easy removal, make cleaning easier
  • Choice of acrylic or stainless-steel covers
View Specifications
Images for Fisher Scientific™ Isotemp™ Digital-Control Water Baths
  • S6307 Series
S6307 Series
S6307 Series
(21 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
15-462-6Q 5L capacity; 5L; Hinged acrylic cover; 230V 60Hz, 2.5A
Accessories
Each for $914.36
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
15-462-28Q 28L capacity; Hinged acrylic cover; 120V 50/60Hz, 8.3A
Accessories
Each for $1,646.00
Check Availability
Add
to Cart
(0) in My Cart
15-462-2SQ 2L capacity; Shallow form; 120V 50/60Hz, 1.7A
Accessories
Each for $841.50
Check Availability
Add
to Cart
(0) in My Cart
15-462-3Q 2L capacity; Hinged acrylic cover; 230V 60Hz, 1.7A
Accessories
Each for $829.72
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
15-462-16Q 5L/10L dual capacity; Hinged acrylic cover; 230V 60Hz, 8.3A
Accessories
Each for $2,101.35
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
15-462-5Q 5L capacity; 120V 50/60Hz, 2.5A
Accessories
Each for $927.50
Check Availability
Add
to Cart
(0) in My Cart
15-462-29Q 28L capacity; Hinged acrylic cover; 230V 60Hz, 8.3A
Accessories
Each for $1,622.60
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
15-462-3SQ 2L capacity; Hinged acrylic cover; 230V 60Hz, 1.7A
Accessories
Each for $829.72
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
15-462-10Q 10L capacity; 120V 50/60Hz, 5.8A; With acrylic cover
Accessories
Each for $1,052.00
Check Availability
Add
to Cart
(0) in My Cart
15-462-2Q 2L capacity; 120V 50/60Hz, 1.7A
Accessories
Each for $841.50
Check Availability
Add
to Cart
(0) in My Cart
15-462-11Q 10L capacity; Hinged acrylic cover; 230V 60Hz, 5.8A
Accessories
Each for $1,037.18
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
15-462-15Q 5L/10L dual capacity; 120V 50/60Hz, 8.3A
Accessories
Each for $2,130.00
Check Availability
Add
to Cart
(0) in My Cart
15-462-20Q 20L capacity; Hinged acrylic cover; 120V 50/60Hz, 8.3A
Accessories
Each for $1,348.00
Check Availability
Add
to Cart
(0) in My Cart
15-462-21Q 20L capacity; Hinged acrylic cover; 230V 60Hz, 8.3A
Accessories
Each for $1,334.12
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
15-462-S15Q 5L/10L dual capacity; Hinged stainless-steel cover; 120V 50/60Hz, 8.3A
Accessories
Each for $3,190.00
Check Availability
Add
to Cart
(0) in My Cart
15-462-S28Q 28L capacity; Hinged stainless-steel cover; 120V 50/60Hz, 8.3A
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
15-462-S20Q 20L capacity; Hinged stainless-steel cover; 120V 50/60Hz, 8.3A
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
15-462-S2Q 2L capacity; Hinged stainless-steel cover; 120V 50/60Hz, 1.7A
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
15-462-S2SQ 2L capacity; Hinged stainless-steel cover; 120V 50/60Hz, 1.7A
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
15-462-S10Q 10L capacity; Hinged stainless-steel cover; 120V 50/60Hz, 5.8A
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
15-462-S5Q 5L capacity; Hinged stainless-steel cover; 120V 50/60Hz, 2.5A
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Add Items to Cart
Description
Microprocessor PID Control
  • ±0.24°C uniformity, ±0.5°C accuracy and stability at 37°C
  • Display shows °C or °F; easy to read from across the room
Safety Features
  • Low profile and front-panel controls for easier access, safer operation
  • Dual thermostats guard against thermal runaway
  • Hinged acrylic or stainless-steel gable covers have fins that keep hands away from hot vapors
  • Covers stay open at 90° or, with quick-release hinge, lift off completely for use with large glassware
  • Heating elements will not burn out if bath accidentally runs dry
Construction
  • Corrosion-resistant exterior with tough, easy-to-clean acrylic enamel coating
  • Temperature-resistant plastic rim insulates, resists leaks, keeps its shape and won't dry out or crack like caulked seals
  • Exterior stays cool even after hours of operation Deep-drawn, stainless-steel reservoirs and perforated sample trays prevent leaks, guard against rust and contamination
  • Rounded reservoir corners and sample trays with finger holes allow for easy removal, make cleaning easier
  • Choice of acrylic or stainless-steel covers
Shallow-form Model 202S
  • Perfect for heating samples in microcentrifuge tubes and for other molecular biology procedures
Dual-Chamber Model 215
  • Features two chambers with separate controls
Models 220 and 228
  • Feature movable control panel that mounts on long or short side of bath
Models 210, 215, 220 and 228
  • With drain pump to empty reservoir
Specifications & Ordering Information:
Additional voltages and models with analog controllers available; contact your Fisher Scientific Customer Service Representative for details.
Certifications: CSA approved.
Warranty: One year
Service agreement available.
Specifications
Temperature Range, Maximum w/LidAmbient to 100°C (212°F)
Temperature Accuracy at 37°C±0.5°C
Temperature Uniformity at 37°C±0.24°C
Temperature Stability at 37°C±0.5°C
Temperature Control/DisplayPID/Digital ft.
Electrical Requirements 120V 50/60Hz
Resolution±0.1°C
ft.Microprocessor PID Control