Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
12-144Q with 12-142T thermometer (order separately)
Fisher-Johns Melting Point Apparatus
Handles materials with melting points from 20° to 300°C
Includes: Apparatus with 0.5g silver wool, 0.5 oz. (14.2g), 0.68 in. (18mm) cover glasses and instructions.
Required Accessories: Mercury thermometer (12-142T)
  • Excellent temperature control and reproducibility (±0.5°C) achieved with variable transformer graduated in percent power
  • Observe melting through convenient, adjustable magnifier
  • Integral lens focuses a light beam on the stage to aid observation
  • Unit is heated with a replaceable 100-watt heating cartridge
  • Unit doubles as a micro-hotplate
  • Handles evaporations and small preparations of organic compounds
  • Base has recess for extra beakers
View Specifications
Images for Fisher-Johns Melting Point Apparatus
  • 12-144Q with 12-142T thermometer (order separately)
12-144Q with 12-142T thermometer (order separately)
12-144Q with 12-142T thermometer (order separately)
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
12-144-1Q For 230V 50/60Hz
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Description
  • Excellent temperature control and reproducibility (±0.5°C) achieved with variable transformer graduated in percent power
  • Observe melting through convenient, adjustable magnifier
  • Integral lens focuses a light beam on the stage to aid observation
  • Unit is heated with a replaceable 100-watt heating cartridge
  • Unit doubles as a micro-hotplate
  • Handles evaporations and small preparations of organic compounds
  • Base has recess for extra beakers
Specifications & Ordering Information:
Size: 9L x 14 in.W (23 x 36cm)
Certifications: CSA approved
Warranty: 12 months
Service agreement available.