Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
22-130-558
Available on GSA/VA Contract for Federal Government customers only. Therapak™ Urine Cytology Collection and Transport Kit
Designed for labs performing urine cytology or FISH testing
Includes: Gender-friendly collection cup, leakproof transport container, urine preservative solution, absorbent pad, patient ID labels, Biological Substance Category B label, and a rigid outer box for return shipment by air or ground.
  • Convenient method for collection, preservation, and transport of urine specimens from off-site locations
  • Easy-to-use—after sample collection, pour 2 oz. of urine into the blue cap container, add 2 oz. of preservative from the green cap vials, tighten the blue cap until it clicks, and prepare for shipment
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Therapak™ Urine Cytology Collection and Transport Kit
  • 22-130-558
22-130-558
22-130-558
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
22-130-055 Available on GSA/VA Contract for Federal Government customers only. 
Therapak Corporation
No.:37928
Urine Cytology Collection and Transport Kit
Description
  • Convenient method for collection, preservation, and transport of urine specimens from off-site locations
  • Easy-to-use—after sample collection, pour 2 oz. of urine into the blue cap container, add 2 oz. of preservative from the green cap vials, tighten the blue cap until it clicks, and prepare for shipment
Compliance: IATA compliant for air or ground shipment; blue cap container is 95kPa certified.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.