Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
View Image Gallery (1) Image(s) are representative of the product Group
  • ricca
Sodium Bicarbonate, Saturated Aqueous Solution, Ricca Chemical
Sodium Bicarbonate
Clear, colorless liquid
Slightly Alkaline
Solvent/Matrix: Saturated Aqueous Solution
[7732-18-5, 89.00-93.00%], [144-55-8, 8.91-9.28%]
Images for Sodium Bicarbonate, Saturated Aqueous Solution, Ricca Chemical
  • ricca
(4 item(s) in product Group)
  • Items
  • Physical Properties
Description Catalog No. Availability Quantity Price
Poly natural; 1L
714832
Ricca Chemical
No.:7148-32
N/AYou must be logged in and have a valid account number to view item availability   Each for $32.80
(0) in My Cart
Cubitainer; 20L
71485
Ricca Chemical
No.:7148-5
N/AYou must be logged in and have a valid account number to view item availability   Each for $181.10
(0) in My Cart
Poly natural; 4L
71481
Ricca Chemical
No.:7148-1
N/AYou must be logged in and have a valid account number to view item availability   Each for $50.70
(0) in My Cart
Poly natural; 500mL
714816
Ricca Chemical
No.:7148-16
N/AYou must be logged in and have a valid account number to view item availability   Each for $22.90
(0) in My Cart
Add Items to Cart
Boiling Point: Approx. 100
Melting Point: Approx. 0
Inifinite
Sp. gr.: Approx. 1 - 1.1