Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
19-165-920
Thermo Scientific™ personal DataRAM™ pDR-1000AN Monitor
Palm-sized unit measures mass concentrations of dust, smoke, mists, and fumes in real time and sounds an audible alarm whenever user-defined level is exceeded
Utilizing single beam nephelometry, the pDR-1000AN incorporates light-scattering to produce optimal particle response for high correlation with standard gravimetric measurements. With datalogging enabled, the pDR-1000AN automatically tags and time stamps the collected data to be stored for subsequent retrieval, printing or computer graphing.
Includes: Universal power supply, PC communications software, zeroing kit, belt clip kit, carrying case, signal output cables, and instruction manual.
Key Features:
  • US MSHA Intrinsic safety approval
  • Easy portability for belt or shoulder strap attachment
  • Flexible data logging routines
  • Audible alarm settings for immediate notification of exceeded user-defined levels
View Specifications
Images for Thermo Scientific™ personal DataRAM™ pDR-1000AN Monitor
  • 19-165-920
19-165-920
19-165-920
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
19165920
THERMO SCI SAFETY
No.:PDR-1000AN
Personal DataRAM pDR-1000AN
Each for $5,379.09
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
Key Features:
  • US MSHA Intrinsic safety approval
  • Easy portability for belt or shoulder strap attachment
  • Flexible data logging routines
  • Audible alarm settings for immediate notification of exceeded user-defined levels
Additional Information:
  • Light-scattering photometer offers high correlation with gravimetric measurement of respirable and thoracic fractions with optimal response to 0.1-10µm size particles
  • Datalogger automatically tags and time stamps data collected and stores it for subsequent retrieval, printing, or graphing through a PC
  • No moving parts means silent operation and minimal maintenance costs
  • Unit is gravimetrically calibrated in mg/m3 and is ready to use after easy zeroing step (kit included)
  • Internal firmware controls automatic calibration check; gravitimetric calibration can be performed in the field for maximum efficiency
  • Concentration Display Updating Interval: 1 second
  • Datalogging Averaging Periods (user selectable): 1 second to 1 hour
  • Total Number of Data Points in Memory: 13,391
  • Number of Data Tags: 99 (maximum)
  • Serial Interface: RS-232, 4800 baud
  • Computer Requirements: PC compatible, 486 processor or higher; Windows™ 95 or higher
Certifications: CE certified. MSHA approved
Compliance: Meets U.S. FCC Rules (Part 15). MSHA intrinsic safety approval (for coal-mining environments containing methane).
DataRAM pDR-1000AN Specifications
Concentration Measurement Ranges (auto ranging)0.001 to 400 mg/m3 (auto ranging)
Scattered Coefficient Range1.5 x 10-6 to 0.6m-1 (approx.) @ lambda = 880nm (not displayed)
Precision/Repeatability Over 30 Days±0.5 of reading or ±0.015 mg/m3, whichever is larger, for 1 second averaging time
±0.5 of reading or ±0.015 mg/m3, whichever is larger for 10 second averaging time
±0.2% of reading or ±0.005 mg/m3, whichever is larger for 60 second averaging time±
Accuracy±5% of reading ± precision (traceable to SAE Fine Test Dust)
Resolution0.1% of reading or 0.001 mg/m3, whichever is larger
Particle Size Range of Max. Response0.1 to 10µm
Aerodynamic Particle Cut-Point Range10µm nominal
Concentration Display Updating Interval1 second
Concentration Display Averaging Time1 to 60 seconds (user selectable)
Data Logging Averaging Periods1 second to 1 hour
Total # of Data Points Logged in Memory13,391
Number of Data Tags99 (maximum)
Logged DataAverage concentration, time/date, and data point number
Readout DisplayLCD 16 characters (4mm height) x 2 lines
Serial InterfaceRS-232 / 4,800 baud
Computer RequirementsIBM-PC compatible, 486 or higher, Windows 95™ or higher, >= 8MB memory, Hard disc drive 3.5 in. floppy, VGA or higher resolution monitor
Analog Output0 to 5V and 4 to 20mA
Physical Dimensions63 x 92 x 153mm (2.5 x 3.6 x 6.0 in.)
Weight0.5kg (18 oz.)