Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Thermo Scientific™ Nalgene™ Wide-Mouth HDPE Sterile Sample Bottle; HDPE, PP screw closure
For water and wastewater samples
  • HDPE container with leak-proof HDPE closure
  • Leak-proof guaranteed
  • Certified Sterile
  • Caps assembled onto containers and individually shrink wrapped for tamper evidence and sterility assurance
  • Packaged in shrink-wrapped trays
  • Wide mouth for easy aseptic filling
  • Recommended for collecting water samples for microbiological analyses
View Specifications
Images for Thermo Scientific™ Nalgene™ Wide-Mouth HDPE Sterile Sample Bottle; HDPE, PP screw closure
(2 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
05-719-359A
Thermo Scientific
No.:N411-0125
125mL WM Nalgene HDPE Bottle Sterile
Case of 48 for $179.52
Check Availability
Add
to Cart
(0) in My Cart
05-719-359B
Thermo Scientific
No.:N411-0250
250mL WM Nalgene HDPE Bottle Sterile
Case of 60 for $284.29
Check Availability
Add
to Cart
(0) in My Cart
Description
  • HDPE container with leak-proof HDPE closure
  • Leak-proof guaranteed
  • Certified Sterile
  • Caps assembled onto containers and individually shrink wrapped for tamper evidence and sterility assurance
  • Packaged in shrink-wrapped trays
  • Wide mouth for easy aseptic filling
  • Recommended for collecting water samples for microbiological analyses
Includes:
Certified sterile containers come with the following features:
  • Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
  • Each container individually barcoded with lot number and unique container number
  • Each closure individually sealed with tamper-evident shrink wrap
  • Analysis labels included unattached for optional use
  • Carton sealed with security tape providing tamper evidence
Compliance: Certified sterile as well as for metals and water quality parameters; manufactured under an ISO certified quality management system.