Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
01-814-50 to -53
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Heat-Shrink Bands for Nalgene™ PETG Media Bottles
Tamper-resistant seal
  • Gamma stable and include a perforated "tear strip" feature for easy removal
  • Available in 4 sizes to mate with Nalgene PETG Square Media Bottles
  • Packed 1,000 per case (two zipper bags of500 each)
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Heat-Shrink Bands for Nalgene™ PETG Media Bottles
  • 01-814-50 to -53
01-814-50 to -53
01-814-50 to -53
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
01-814-50 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:312160-0200
Heat-shrink band for 30mL PETG Square Media Bottle with 24-415 HDPE Closure
Case of 1000 for $147.00
Check Availability
Add
to Cart
(0) in My Cart
01-814-51 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:312160-0240
Heat-shrink band for 60mL PETG Square Media Bottle with 24-415 HDPE Closure
Case of 1000 for $174.30
Check Availability
Add
to Cart
(0) in My Cart
01-814-52 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:312160-0384
Heat-shrink band for 125-1000mL PETG Square Media Bottle with 38-430 HDPE Closure
Case of 1000 for $204.00
Check Availability
Add
to Cart
(0) in My Cart
01-814-53 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:312160-0530
Heat-shrink band for 2000mL PETG Square Media Bottle with 53B HDPE Closure
Case of 1000 for $258.50
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Gamma stable and include a perforated "tear strip" feature for easy removal
  • Available in 4 sizes to mate with Nalgene PETG Square Media Bottles
  • Packed 1,000 per case (two zipper bags of500 each)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.