Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Acetate Buffer Solution, pH 3.5, For USP Heavy Metals Testing, Ricca Chemical™
Acetate Buffer Solution
Clear, colorless liquid
For USP Heavy Metals Testing
pH 3.5
[7732-18-5, 63.00-66.00%], [7647-01-0, 18.71-19.47%], [631-61-8, 15.87-16.51%]
Images for Acetate Buffer Solution, pH 3.5, For USP Heavy Metals Testing, Ricca Chemical™
  • ricca
(1 item(s) in product Group)
  • Items
  • Physical Properties
  • Safety and Handling
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
60132
Ricca Chemical
No.:60.1-32
1L; Poly natural
MSDS
Each for $191.50
Check Availability
Add
to Cart
(0) in My Cart
Boiling Point: Approx. 100-118
Melting Point: Approx. 0-16.6
Shelf life: 24 Months