Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
AB-1850
View Image Gallery (2) Image(s) are representative of the product Group
  • AB-1850
  • AB-1850
Thermo Scientific™ VisionMate™ High Speed 2D Barcode Reader
Camera based compact unit for high speed scanning of 2D barcoded tubes in a cluster rack
Speed and Versatility
  • Intuitive software can be easily downloaded for manual use or remote control through TCP/IP connection
  • Scans a single 2D barcoded storage tube or entire tube rack in less than one second
  • Robust optics and compact, integration-friendly design are ideal for high throughput use
Enhanced Tube Readability
  • Configured for all Thermo Scientific 2D barcoded tubes and user configurable for other 2D tube formats
  • Integration of linear 1D barcode reader allows integration of individual tube and rack barcode data
Simple Operation
  • Connect to your computer via a single USB port and, despite which way the tube rack is placed, the computer automatically adjusts orientation and correctly aligns codes with well positions
  • Connects to PC via a single USB port of easy installation
  • Single camera enables save of image file
  • Automatic rack orientation detection and adjustment of 2D barcode read data
Superior Sample Protection
  • Ability to operate down to -40°C enables barcode reading in cold rooms and low temperature liquid stores to preserve sample integrity
  • Sealed unit prevents frost and condensation which allows reading of frozen tubes at ambient temperatures
Data File Flexibility
  • Easily integrated into LIMS, databases, or tracking systems
  • User-defined code types and lengths indicate incorrect codes and limit access by user profile
  • Optimized software allows users the ability to select the export data format and conserve memory space
View Specifications
Images for Thermo Scientific™ VisionMate™ High Speed 2D Barcode Reader
  • AB-1850
  • AB-1850
AB-1850
AB-1850
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
AB1850
Thermo Scientific
No.:AB-1850
VisionMate High Speed 2D Barcode Reader
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Speed and Versatility
  • Intuitive software can be easily downloaded for manual use or remote control through TCP/IP connection
  • Scans a single 2D barcoded storage tube or entire tube rack in less than one second
  • Robust optics and compact, integration-friendly design are ideal for high throughput use
Enhanced Tube Readability
  • Configured for all Thermo Scientific 2D barcoded tubes and user configurable for other 2D tube formats
  • Integration of linear 1D barcode reader allows integration of individual tube and rack barcode data
Simple Operation
  • Connect to your computer via a single USB port and, despite which way the tube rack is placed, the computer automatically adjusts orientation and correctly aligns codes with well positions
  • Connects to PC via a single USB port of easy installation
  • Single camera enables save of image file
  • Automatic rack orientation detection and adjustment of 2D barcode read data
Superior Sample Protection
  • Ability to operate down to -40°C enables barcode reading in cold rooms and low temperature liquid stores to preserve sample integrity
  • Sealed unit prevents frost and condensation which allows reading of frozen tubes at ambient temperatures
Data File Flexibility
  • Easily integrated into LIMS, databases, or tracking systems
  • User-defined code types and lengths indicate incorrect codes and limit access by user profile
  • Optimized software allows users the ability to select the export data format and conserve memory space
Specifications
Speed< 1 second/rack
Decoding Capacity1 rack
Data Transfer MethodData transfer to comma-delimited linear and columnar ASCII text files; ActiveX controls
Dimensions (D x W x H)8 x 8.75 x 7.25 in. (20.5 x 22.5 x 18.5cm)
Weight5 lbs. (2.27kg)
Power Supply115-240 V AC, 50-60 Hz
Supported BarcodeAny Thermo Scientific Matrix 2D Barcoded Storage Tube in 8x12 or 4x6 format as well as virtually any other 2D barcode in similar formats
Computer RequirementsMicrosoft™ Windows™ controller PC