Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
03-421-209 to -212
Available on GSA/VA Contract for Federal Government customers only. Hamilton Scientific™ Concept™ Bypass Fume Hoods with Vertical Frameless Sash
Feature 35 in. (89cm) sash opening for optimal visibility
Available pre-wired and pre-plumbed (4-6 weeks additional lead time).
Required Accessories: Ductwork, motor blower, service fixtures, work surface, base cabinets.
  • Exhaust volumes from 484 to 1084cfm (0.09 to 0.15H2O), depending on hood size
  • Designed for 60 to 100fpm face velocities
  • Downdraft bypass for increased volume, less resistance, quieter operation
  • Low-profile airfoil provides obstruction-free access to the hood interior
  • Frameless vertical rising sash
  • Chain and sprocket sash support with alignment shaft and sash leveling system
  • Fixed-baffle system requires no adjustment
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Hamilton Scientific™ Concept™ Bypass Fume Hoods with Vertical Frameless Sash
  • 03-421-209 to -212
03-421-209 to -212
03-421-209 to -212
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
03-421-209 Available on GSA/VA Contract for Federal Government customers only. 
Hamilton Scientific
No.:HM54L2570P0B
48 in. hood; Total exhaust pressure loss: 484cfm
Each for $7,370.00
Check Availability
Add
to Cart
(0) in My Cart
03-421-210 Available on GSA/VA Contract for Federal Government customers only. 
Hamilton Scientific
No.:HM54L2571P0B
60 in. hood; Total exhaust pressure loss: 634cfm
Each for $7,670.00
Check Availability
Add
to Cart
(0) in My Cart
03-421-211 Available on GSA/VA Contract for Federal Government customers only. 
Hamilton Scientific
No.:HM54L2572P0B
72 in. hood; Total exhaust pressure loss: 784cfm
Each for $8,410.00
Check Availability
Add
to Cart
(0) in My Cart
03-421-213Available on GSA/VA Contract for Federal Government customers only. 
Hamilton Scientific
No.:HM54L2574P0B
96 in. hood; Total exhaust pressure loss: 1084cfm
Each for $9,810.00
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Exhaust volumes from 484 to 1084cfm (0.09 to 0.15H2O), depending on hood size
  • Designed for 60 to 100fpm face velocities
  • Downdraft bypass for increased volume, less resistance, quieter operation
  • Low-profile airfoil provides obstruction-free access to the hood interior
  • Frameless vertical rising sash
  • Chain and sprocket sash support with alignment shaft and sash leveling system
  • Fixed-baffle system requires no adjustment
Safety Features
  • Secondary trough provides an additional safeguard for the containment of spills AutoSash™ locks in the setup position, automatically returns to maximum operational height when released
  • Construction
    • White polyresin lining
    • Side wall access panels with PVC gasket
    • Plugged holes for fixture installation on both posts
    • Two tube fluorescent light fixtures
    • Light switch
    • Two duplex 120V electrical receptacles
    • Overall 31.25D x 63.5 in.H (79 x 161cm)
    Specifications & Ordering Information:
    Supplied without motor blower, service fixtures, work surface or base cabinets for support. Installation by a certified Thermo Scientific installer recommended.
    Certifications: UL 1805 Classified, ASHRAE™ 110 Tested
    Note: Not for use with standard Hamilton Scientific work surfaces.
    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.