Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
UFV5 Series
EMD Millipore Ultrafree™ Centrifugal Filter Devices†
Rapid concentration, desalting and purification in as little as 10 minutes
Combine the design of a centrifugal filter device with a vertically oriented membrane, for quick and efficient sample processing.
  • Polycarbonate reservoir
  • Concentrate, purify and prepare biological samples for volumes up to 0.5mL
  • High-flow, Biomax™ ultrafiltration membrane available in NMWL cutoffs
  • Recover processed sample from concentrate pocket after a single spin
  • Depending on sample type, concentration factors range from 50x to 200x
  • Can be used with any fixed-angle or swinging bucket centrifuge with standard rotors which accept standard receiver tubes
View Specifications
Images for EMD Millipore Ultrafree™ Centrifugal Filter Devices†
  • UFV5 Series
UFV5 Series
UFV5 Series
(2 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
UFV5 BTK 25
EMD Millipore
No.:UFV5BTK25
NMWL: 30,000; 500µL; 25/Pk.
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
UFV5 BHK 25
EMD Millipore
No.:UFV5BHK25
NMWL: 100,000; 500µL; 25/Pk.
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
  • Polycarbonate reservoir
  • Concentrate, purify and prepare biological samples for volumes up to 0.5mL
  • High-flow, Biomax™ ultrafiltration membrane available in NMWL cutoffs
  • Recover processed sample from concentrate pocket after a single spin
  • Depending on sample type, concentration factors range from 50x to 200x
  • Can be used with any fixed-angle or swinging bucket centrifuge with standard rotors which accept standard receiver tubes
Ultrafree-0.5
Max. Volume Capacity0.5mL
Minimum Final Concentrate Volume5µL
Max. Centrifugal Force12,000xG (10,000xG for Biomax-100)
Active Membrane Surface Area0.48cm2
Hold-Up Volume of Membrane<8µL
Dia. x Length7/16 x 2 in. (10.7 x 50mm)
RotorFixed angle or swinging bucket
†An Amicon™ Bioseparations product.