Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
View Image Gallery (2) Image(s) are representative of the product Group
  • acros
Available on GSA/VA Contract for Federal Government customers only. Methyl sulfoxide, 99.7+%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™
Methyl sulfoxide
Extra Dry over Molecular Sieve
99.7+%
Dimethyl sulfoxide, DMSO
C2H6OS
(CH3)2SO
F.W.: 78.13
[67-68-5]
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Methyl sulfoxide, 99.7+%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™
  • acros
(4 item(s) in product Group)
  • Items
  • Specifications
  • Physical Properties
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
AC348445000
ACROS Organics
No.:348445000
500mL; AcroSeal glass bottle
MSDS
Each for $72.20
Check Availability
Add
to Cart
(0) in My Cart
AC34844-0010 Available on GSA/VA Contract for Federal Government customers only. 
ACROS Organics
No.:348440010
1L; AcroSeal glass bottle
MSDS
Each for $102.80
N/A N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
AC348440025
ACROS Organics
No.:348440025
2.5L; AcroSeal glass bottle
MSDS
Each for $230.80
N/A N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
AC34844-1000 Available on GSA/VA Contract for Federal Government customers only. 
ACROS Organics
No.:348441000
100mL; AcroSeal glass bottle
MSDS
Each for $54.30
N/A N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
Product Specifications
Assay99.70 % min. (GC)
Color5 APHA max.
Acetic Acid0.003% max.
Physical FormWhite to yellow to brown powder, needles or crystals
Infrared SpectrumAuthentic
Water0.005 % max.
Additional InformationVapor Pressure: 0.55mbar at 20°C
Boiling Point: 189.0°C
Decomposition Information: >190°C
Density: 1.1000g/mL
Flash Point: 87°C
Melting Point: 18.4°C
n: 1.4770 to 1.4800
Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
Sp. gr.: 1.1000
1.987 mPa.s (25°C)