Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
22-046 Series
Available on GSA/VA Contract for Federal Government customers only. Polymedco™ SED-CHEK 2™ ESR Controls
Unique cell-stabilizing process mimics the rouleaux formation of patient samples
  • Bi-level, whole-blood control designed to monitor ESR procedures
  • Monitor technique as well as environmental and physical factors that adversely affect ESR results
  • Run like a patient sample—not necessary to void diluent from pre-filled tubes
  • Universal control assayed for both automated and manual ESR methods
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Polymedco™ SED-CHEK 2™ ESR Controls
  • 22-046 Series
22-046 Series
22-046 Series
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
22-046-911 Available on GSA/VA Contract for Federal Government customers only. 
Polymedco
No.:ESR-2CT
6 x 8mL
Each for $362.92
Each for $344.77 Online Instant Savings (5%)
Check Availability
Add
to Cart
(0) in My Cart
22-046-912 Available on GSA/VA Contract for Federal Government customers only. 
Polymedco
No.:ESR2CTA
Abnormal Level; 6 x 8mL
Each for $375.42
Each for $356.65 Online Instant Savings (5%)
Check Availability
Add
to Cart
(0) in My Cart
22-046-913 Available on GSA/VA Contract for Federal Government customers only. 
Polymedco
No.:ESR2CTN
Normal Level; 6 x 8mL
Each for $375.42
Each for $356.65 Online Instant Savings (5%)
Check Availability
Add
to Cart
(0) in My Cart
22-469-17 Available on GSA/VA Contract for Federal Government customers only. 
Polymedco
No.:ESR-5CT
6 x 8mL; 3 abnormal and 3 normal
Pack for $368.75
Pack for $350.31 Online Instant Savings (5%)
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Bi-level, whole-blood control designed to monitor ESR procedures
  • Monitor technique as well as environmental and physical factors that adversely affect ESR results
  • Run like a patient sample—not necessary to void diluent from pre-filled tubes
  • Universal control assayed for both automated and manual ESR methods
Specifications & Ordering Information:
Controls packaged in 6 x 8mL vials
Recommended Storage: 18-month closed vial stability from date of manufacture. 31-day open vial stability. Store at room temperature.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.