Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
02292 Series
View Image Gallery (1) Image(s) are representative of the product Group
  • 02292 Series
Available on GSA/VA Contract for Federal Government customers only. Fisher Diagnostics* Pacific Hemostasis* Activated Partial Thromboplastin Time (aPTT) Reagents
aPTT reagents provide sensitivity to intrinsic and common pathway factors, heparin and lupus anticoagulant
Pacific Hemostasis Activated Partial Thromboplastin Time reagents (APTT) are used to identify acquired or congenital disorders to the intrinsic and common pathways. APTT testing is widely accepted as a means of monitoring patients on oral anticoagulant therapy such as heparin.
  • 10mL vials can complete ~1000 full volume, single determinations; 4mL vials can complete ~400 full volume, single determinations
  • Open vial stability: 30 days at 2° to 8°C
aPTT-XL (Ellagic Acid Activator) (Product No. 022929)
  • Superior factor sensitivity and responsiveness
  • Increased sensitivity to heparin and lupus-like inhibitors
KONTACT*(Particulate Activator) (Product No. 022924)
  • Higher sensitivity to Factor VIII.
  • Average Normal Values:  26 - 33 Seconds
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Fisher Diagnostics* Pacific Hemostasis* Activated Partial Thromboplastin Time (aPTT) Reagents
  • 02292 Series
02292 Series
(7 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
KONTACT (Micronized Silica Activator) 10 x 10mL8
022924 N/A N/A N/A
This product is regulated by the federal government, and may not be ordered online.  This is a National Order Desk item, and cannot be ordered online. 
CaCl2, 0.02M, 10 x 10mL5
MSDS
285545 Available on GSA/VA Contract for Federal Government customers only.  N/A N/A N/A
This product is regulated by the federal government, and may not be ordered online.  This is a National Order Desk item, and cannot be ordered online. 
CaCl2, 0.025M, 10 x 4mL
MSDS
022925 N/A N/A N/A
This product is regulated by the federal government, and may not be ordered online.  This is a National Order Desk item, and cannot be ordered online. 
aPTT-XL, 10 x 10mL
022927 Available on GSA/VA Contract for Federal Government customers only.  N/A N/A N/A
This product is regulated by the federal government, and may not be ordered online.  This is a National Order Desk item, and cannot be ordered online. 
CaCl2, 0.025M, 10 x 10mL
MSDS
176149 N/A N/A N/A
This product is regulated by the federal government, and may not be ordered online.  This is a National Order Desk item, and cannot be ordered online. 
APTT-XL (Ellagic Acid Activator) 10 x 4mL
022929 N/A N/A N/A
This product is regulated by the federal government, and may not be ordered online.  This is a National Order Desk item, and cannot be ordered online. 
aPTT-LS, 10 x 10mL
MSDS
022926 Available on GSA/VA Contract for Federal Government customers only.  N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This is a National Order Desk item, and cannot be ordered online.
View substitute products »
Add Items to Cart
Description
  • 10mL vials can complete ~1000 full volume, single determinations; 4mL vials can complete ~400 full volume, single determinations
  • Open vial stability: 30 days at 2° to 8°C
aPTT-XL (Ellagic Acid Activator) (Product No. 022929)
  • Superior factor sensitivity and responsiveness
  • Increased sensitivity to heparin and lupus-like inhibitors
KONTACT*(Particulate Activator) (Product No. 022924)
  • Higher sensitivity to Factor VIII.
  • Average Normal Values:  26 - 33 Seconds
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.