Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
05-724-50
View Image Gallery (1) Image(s) are representative of the product Group
  • 05-724-50
Chempure* HPLC Solvent Filter/Degasser
For routine HPLC filtration of up to 4L of liquid. Filters out suspended particles, degasses solvents. Attaches to standard bottles and flasks via 38mm adapter cap. Vacuum hose attaches to side tube; all components in contact with filtrate are PTFE. System uses 47mm diameter (0.2µm) PTFE filter. Resists strong acids and bases; suitable for use with most aqueous and organic solvents.
View Specifications
Images for Chempure* HPLC Solvent Filter/Degasser
  • 05-724-50
05-724-50
(1 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
Filter Degasser
05-724-50
Lazar Research Laboratories
No.:FG-256
N/AYou must be logged in and have a valid account number to view item availability   Each for $1,188.00
(0) in My Cart
Description
For routine HPLC filtration of up to 4L of liquid. Filters out suspended particles, degasses solvents. Attaches to standard bottles and flasks via 38mm adapter cap. Vacuum hose attaches to side tube; all components in contact with filtrate are PTFE. System uses 47mm diameter (0.2µm) PTFE filter. Resists strong acids and bases; suitable for use with most aqueous and organic solvents.