Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
09-740-105 to -120
View Image Gallery (2) Image(s) are representative of the product Group
  • 09-740-105 to -120
  • 09-740-105 to -120
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Nalgene™ Syringe Filters - 25mm Diameter
Ideal for filtration of 10 to 100mL volumes
A broad choice of membranes and pore sizes are available for sterile and non-sterile laboratory applications with aqueous and non-aqueous fluids, as well as gases. Nalgene 25mm diameter syringe filters perform as well as 30mm or 33mm diameter products† in most applications, but produce less plastic waste.
  • 100 to 100mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip
  • Polypropylene housing with ID color rings
  • Pore size and membrane type printed on unit
Polyethersulfone (PES)
  • Lowest protein binding and extractables
  • Best choice for filtering cell culture media
  • Polypropylene housings with green color ring
Surfactant-free Cellulose Acetate (SFCA)
  • Low protein binding, lower extractables than standard CA
  • Good for cell culture fluids and cellular solutions
  • Polypropylene housings with blue color rings
  • Membrane of choice for sterilization
Cellulose Acetate (CA)
  • Some protein binding and extractables
  • For sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Polypropylene housings with red color rings
Nylon
  • Naturally hydrophilic, low extractables, some protein binding
  • Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
  • Polypropylene housings with pink color rings
PTFE
  • Naturally hydrophobic
  • Excellent for HPLC/GC solvent filtration; ideal for sterile venting of air or other gases during filtration
  • Polypropylene housings with white color ring
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Nalgene™ Syringe Filters - 25mm Diameter
  • 09-740-105 to -120
  • 09-740-105 to -120
09-740-105 to -120
09-740-105 to -120
(32 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
09-740-120
Thermo Scientific
No.:728-2045
PTFE; 0.45µm; Nonsterile; 50/Pk.
Case of 500 for $1,810.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-108
Thermo Scientific
No.:7239920
SFCA; 0.2µm pore; Sterile; 125/pk.
Case of 125 for $398.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-105
Thermo Scientific
No.:723-2520
SFCA; 0.2um pore; Sterile
Case of 50 for $163.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-111
NNI
No.:724-2045
SFCA; 0.45µm Nonsterile
Case of 500 for $1,550.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-35D
NNI
No.:191-2020
FILT SYR BLK NONST.20UM 50/PK
Pack of 50 for $285.43
Case of 6 PK for $1,005.10
Check Availability
Add
to Cart
(0) in My Cart
09-740-35J
Thermo Scientific
No.:195-2520
FILTER SYRINGE .20UM 50/CS
Case of 50 for $224.12
Check Availability
Add
to Cart
(0) in My Cart
09-740-112
NNI
No.:7242080
CA; 0.8µm pore; Nonsterile; 50/pk.
Case of 500 for $1,550.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-114
NNI
No.:7252545
PES; Pore Size: 0.45µm; Sterile
Case of 50 for $163.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-37G
Thermo Scientific
No.:1909945
SYR FIL BK SFCA .45 125/CS
Case of 125 for $443.40
Check Availability
Add
to Cart
(0) in My Cart
09-740-113
NNI
No.:725-2520
PES; Pore Size: 0.2µm; Sterile
Case of 50 for $163.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-35F
NNI
No.:191-2080
FILT SYR BLK N-STL.80UM 50/PK
Pack of 50 for $283.33
Case of 6 PK for $1,008.93
Check Availability
Add
to Cart
(0) in My Cart
09-740-109
Thermo Scientific
No.:7239945
SFCA; 0.45µm pore; Sterile; 125/pk.
Case of 125 for $398.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-110
NNI
No.:7242020
SFCA; 0.2µm Pore; Nonsterile
Case of 500 for $1,550.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-119
Thermo Scientific
No.:7282020
PTFE; 0.2µm pore; Nonsterile
Case of 500 for $1,810.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-118
Thermo Scientific
No.:7272045
Nylon; 0.45µm; Nonsterile
Case of 500 for $1,550.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-107
Thermo Scientific
No.:7232580
CA; 0.8µm pore; Sterile; 50/cs.
Case of 50 for $163.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-117
Thermo Scientific
No.:727-2020
Nylon; 0.2µm pore; Nonsterile
Case of 500 for $1,550.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-106
Thermo Scientific
No.:7232545
SFCA; 0.45um pore; Sterile
Case of 50 for $163.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-38A
Thermo Scientific
No.:199-2020
FILTER SYRNG NONST.20UM 50/PK
Pack of 50 for $336.20
Case of 6 PK for $1,172.53
Check Availability
Add
to Cart
(0) in My Cart
09-740-115
Thermo Scientific
No.:7262520
Nylon; 0.2µm pore; Sterile
Case of 50 for $198.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-116
Thermo Scientific
No.:7262545
Nylon; 0.45µm pore; Sterile
Case of 50 for $198.00
Check Availability
Add
to Cart
(0) in My Cart
09-740-35E
NNI
No.:1912045
FILT SYR BLK NONST.45UM 50/PK
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
09-740-37F
Thermo Scientific
No.:190-9920
SYR FIL BK SFCA .2 125/CS
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
09-740-35P
Thermo Scientific
No.:196-2020
FILTER SYRNGE BLK .20UM 50/PK
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
09-740-38B
Thermo Scientific
No.:199-2045
FILTER SYRNG NONST.45UM 50/PK
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
09-740-35Q
Thermo Scientific
No.:196-2045
FILTER SYRNGE BLK .45UM 50/PK
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
09-740-61A
NNI
No.:1942520
FLTR SYR PES 25MM .2UM 50/CS
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
09-740-35B
Thermo Scientific
No.:190-2545
FILTER SYRINGE .45UM 50/CS
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
09-740-61B
NNI
No.:1942545
FLTR SYR PES 25MM .45UM 50/CS
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
09-740-35K
Thermo Scientific
No.:195-2545
FILTER SYRINGE .45UM 50/CS
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
09-740-35A
Thermo Scientific
No.:190-2520
FILTER SYRINGE .20UM 50/CS
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
09-740-35C
Thermo Scientific
No.:190-2580
FILTER SYRNG .80UM 25MM 50/CS
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Add Items to Cart
Description
  • 100 to 100mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip
  • Polypropylene housing with ID color rings
  • Pore size and membrane type printed on unit
Polyethersulfone (PES)
  • Lowest protein binding and extractables
  • Best choice for filtering cell culture media
  • Polypropylene housings with green color ring
Surfactant-free Cellulose Acetate (SFCA)
  • Low protein binding, lower extractables than standard CA
  • Good for cell culture fluids and cellular solutions
  • Polypropylene housings with blue color rings
  • Membrane of choice for sterilization
Cellulose Acetate (CA)
  • Some protein binding and extractables
  • For sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Polypropylene housings with red color rings
Nylon
  • Naturally hydrophilic, low extractables, some protein binding
  • Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
  • Polypropylene housings with pink color rings
PTFE
  • Naturally hydrophobic
  • Excellent for HPLC/GC solvent filtration; ideal for sterile venting of air or other gases during filtration
  • Polypropylene housings with white color ring
† Based on the dry volume of 100 syringe filters measured in a 1000mL beaker
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.