Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Lifescan SureStep™ Pro System Test Strips
SureStep Pro Test Strips are for use with SureStep Brand Blood Glucose Meters for quantitatively measuring glucose levels in whole blood.
SureStep Pro Test Strips may be used to test capillary, venous, arterial, and neonatal blood samples.
Required Accessories:
  • SureStep Brand Blood Glucose Meter and applicable operator's guide.
  • Single–use lancing device, such as Lifescan Unisik' 2 or equivalent (If obtaining capillary sample)
  • The test employs a dry reagent technology based on the glucose oxidase method which is specific for D–glucose.
  • When a small drop of whole blood is applied to a SureStep Pro Test Strip, glucose oxidase on the test strip triggers the oxidation of glucose in the blood sample.
  • Peroxidase on the test strip then causes the hydrogen peroxide to react with dyes to produce a blue color in the presence of oxygen.
  • This blue color is visible through the confirmation dot on the back of the test strip—the darker the blue, the higher the glucose level.
  • SureStep Brand Blood Glucose Meters measures the color intensity of the confirmation dot and report a plasma–calibrated glucose result.
View Specifications
Images for Lifescan SureStep™ Pro System Test Strips
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
23-029296
Lifescan
No.:53885079750
Test Strips for SureStep meter
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Description
  • The test employs a dry reagent technology based on the glucose oxidase method which is specific for D–glucose.
  • When a small drop of whole blood is applied to a SureStep Pro Test Strip, glucose oxidase on the test strip triggers the oxidation of glucose in the blood sample.
  • Peroxidase on the test strip then causes the hydrogen peroxide to react with dyes to produce a blue color in the presence of oxygen.
  • This blue color is visible through the confirmation dot on the back of the test strip—the darker the blue, the higher the glucose level.
  • SureStep Brand Blood Glucose Meters measures the color intensity of the confirmation dot and report a plasma–calibrated glucose result.
Recommended Storage: Store test strips in their original bottle in a cool, dry place 30°C (86°F). Keep away from heat and direct sunlight. Do not refrigerate or freeze.