Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Fisher Scientific- null - 610599 - Product Details
Thermo Scientific™ MAS™ Liquid Urinalysis Control
Combines dipstick methods with microscopic components
  • Qualitative/semi-quantitative
  • Values for microalbumin and creatinine on the Bayer™ Clinitek™ 50 and 100 analyzers
  • Additional values for all of the major reagent strips and strip systems
  • 30-day open-vial stability for all analytes when stored at 2° to 8°C
View Specifications
Images for Thermo Scientific™ MAS™ Liquid Urinalysis Control
Description
  • Qualitative/semi-quantitative
  • Values for microalbumin and creatinine on the Bayer™ Clinitek™ 50 and 100 analyzers
  • Additional values for all of the major reagent strips and strip systems
  • 30-day open-vial stability for all analytes when stored at 2° to 8°C
  • "/>
    Safety Features: Applications:
    Specifications & Ordering Information:
    Certifications:
    Compliance:
    Recommended Storage:
    Warranty:
    Liquid Urinalysis Control
    AnalyteUnitsLevel 1Level 2Level 3
    AppearanceClearClearClear
    ColorAmberAmberYellow
    Bilirubin (BMD)2+ to 3+2+ to 3+Negative
    Blood (BMD)Ery/µL25050Negative
    Creatininemg/dL200–30010–5010–50
    CrystalsPresentPresentAbsent
    Glucose (BMD)g/dL0.5–10.25–0.5Normal
    hCGPositiveNegative
    Ketones (BMD)2+ to 3+2+ to 3+Negative
    Leukocytes Esterase (BMD)1+ to 2+1+ to 2+Negative
    Microalbuminmg/L>=15030–8010
    NitritePositivePositiveNegative
    OsmolalitymOsm1000–1200600–800400–600
    pH7.5–8.57.0–8.06.0–7.0
    PotassiummEq/L10.8–11.84.2–5.220.9–21.9
    Protein (BMD)mg/dL100–500Trace–30Negative
    Red Blood Cellsper µL55–8520–600
    SodiummEq/L160–18060–180295–315
    Specific Gravity (Refractometer)1.027–1.0331.017–1.0231.011–1.015
    Urobilinogen (BMD)mg/dL8–12NormalNormal
    White Blood Cellsper µL65–9510–351–10
    "/>
    Refer to package insert for specific analyte and stability claims.