Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Revco High-Performance Lab Freezers
View Image Gallery (2) Image(s) are representative of the product Group
  • Revco High-Performance Lab Freezers
  • 13-990-266, 14-259-233
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Revco™ High-Performance Lab Freezers
Microprocessor control simplifies operation, delivers greater accuracy
  • Temperature factory-preset at -20° or -30°C
  • Capacities from 4.9 to 51.1 cu. ft. (139 to 1447L)
  • Icon-based controls and displays
  • Large, easy-to-read digital display shows temperature within 1°C
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature
Microprocessor Control
  • Audible/visual warnings for overtemperature, undertemperature, or power failure
  • Key-operated OFF/ON/ALARM ON master switch for setpoint security
  • Push-button alarm test
  • Remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Controller battery backup system
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Revco™ High-Performance Lab Freezers
  • Revco High-Performance Lab Freezers
  • 13-990-266, 14-259-233
Revco High-Performance Lab Freezers
Revco High-Performance Lab Freezers
(14 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
13-990-266 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:ULT430A
Automatic defrost; -30deg.C; 4.9 cu. ft.; 115V, 60Hz
Accessories
14-259-233
Thermo Scientific
No.:ULT430D
Automatic defrost; -30deg.C; 4.9 cu. ft.; NEMA 6-15P plug type; 208/230V, 60Hz
22-285-655 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:ULT1230A
Automatic defrost; -30°C; 11.5cf; 115V 60Hz
Accessories
13-990-70 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:ULT2330A
Automatic defrost; -30°C; 23.3 cu. ft.; 115V 60Hz
Accessories
13-989-229 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:ULT2330D
Automatic defrost; -30°C; 23.3 cu. ft.; 208/230V 60Hz
Accessories
13-991-127 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:ULT3030A
Automatic defrost; -30°C; 29.2cf; 115V 60Hz
Accessories
13-989-230 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:ULT3030D
Automatic defrost; -30°C; 29.2 cu. ft.; 208/230V 60Hz
Accessories
13-991-129 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:ULT3030V
Automatic defrost; -30°C; 29.2 cu. ft.; 230V 50Hz
Accessories
22-285-651 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:ULT5030A
Automatic defrost; -30°C; 51.1 cu. ft.; 115V 60Hz
Accessories
22-285-652 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:ULT5030D
Automatic defrost; -30°C; 51.1 cu. ft.; 208/230V 60Hz
Accessories
13-990-205 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:UGL2320A
Manual defrost; -20°C; 23.3cf.; 115V 60Hz
Accessories
13-990-207 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:UGL2320D
Manual defrost; -20°C; 23.3cf; 208/230V 60Hz
Accessories
UGL3020A
Thermo Scientific
No.:UGL3020A
Manual defrost; -20°C; 29.2cf; 115V 60Hz
UGL3020D
Thermo Scientific
No.:UGL3020D
Manual defrost; -20°C; 29.2 cu. ft.; 208/230V 60Hz
Description
  • Temperature factory-preset at -20° or -30°C
  • Capacities from 4.9 to 51.1 cu. ft. (139 to 1447L)
  • Icon-based controls and displays
  • Large, easy-to-read digital display shows temperature within 1°C
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature
Microprocessor Control
  • Audible/visual warnings for overtemperature, undertemperature, or power failure
  • Key-operated OFF/ON/ALARM ON master switch for setpoint security
  • Push-button alarm test
  • Remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Controller battery backup system
Construction
  • Scratch-resistant cold-rolled steel interior and exterior finishes
  • High-density, 2 in. thick (5.1cm) CFC-free urethane foam insulation protects against ambient temperature fluctuations
  • CFC-free, HFC-blend refrigerant
  • Single-door models are hinged on right
  • Full-length door handle and key lock
  • Doors self-close and lock open at 90°
  • Adjustable epoxy-coated wire shelves
  • 1 in. (2.5cm) dia. access port with plug on undercounter model
-30°C Freezers
  • Automatic defrost and condensate removal
  • Sensor automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost buildup, and optimizing coil efficiency
  • Forced-air circulation improves temperature uniformity, provides faster recovery after door is opened
-20°C Freezers
  • Suitable for applications in which slight intermittent coil warming during an automatic defrost cycle is not desired (such as enzyme storage)
  • Gravity convection air system for maximum stability without a blower or fan
  • Manual defrost
Specifications & Ordering Information:
Alarm systems, temperature chart recorders, and power monitors are available separately.
Compliance: UL and CUL listed. CE marked.
Warranty: Two years parts and labor (except 13-991-129, which is 16 months parts and labor)
Specifications
Temperature ControlMicroprocessor
RefrigerantCFC-free
InsulationCFC-free urethane foam
CabinetScratch-resistant painted steel
ChamberScratch-resistant painted steel
Note: Published interior volumes are nominal; usable capacities may be offset by shelves, drawers, blower coils or cabinet protrusions. For specific cabinet dimensions and allowances, or for verification of clearance tolerances, contact your Customer Service Representative
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.