Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
13-678-45
View Image Gallery (4) Image(s) are representative of the product Group
  • 13-678-45
  • 13-678-45
  • 13-678-45
  • 13-678-45
Available on GSA/VA Contract for Federal Government customers only. Roche™ Diagnostics ACCU-CHEK™ Safe-T-Pro™ Lancets
Designed with Safe-T-Wings tabs for patient and user protection from reuse or accidental contact
  • Single-use, disposable lancet for taking fingerstick samples of whole blood
  • T-shape makes handling easier
  • When blue trigger button is pressed to obtain a sample, spring-loaded firing action breaks the lancet wings to provide a superior fail-safe against reuse
  • For added protection, lancet permanently retracts into its protective case after use to prevent accidental needlesticks and contamination
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Roche™ Diagnostics ACCU-CHEK™ Safe-T-Pro™ Lancets
  • 13-678-45
  • 13-678-45
  • 13-678-45
  • 13-678-45
13-678-45
13-678-45
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
13-678-45 Available on GSA/VA Contract for Federal Government customers only. 
Roche
No.:03136752001
Safe-T-Pro Lancets
Pack of 200 for $124.71
Pack of 200 for $106.00 Online Instant Savings (15%)
Case of 6 PK for $631.44
Case of 6 PK for $536.72 Online Instant Savings (15%)
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Single-use, disposable lancet for taking fingerstick samples of whole blood
  • T-shape makes handling easier
  • When blue trigger button is pressed to obtain a sample, spring-loaded firing action breaks the lancet wings to provide a superior fail-safe against reuse
  • For added protection, lancet permanently retracts into its protective case after use to prevent accidental needlesticks and contamination
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.