Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
13-676-10M
View Image Gallery (3) Image(s) are representative of the product Group
  • 13-676-10M
  • 13-678-11 Series pipets have color-coded, transparent paper/plastic wrappers for easy identification and tear-through.
  • 13-676-10 and 13-678-11 Series
Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Sterile Polystyrene Disposable Serological Pipettes with Magnifier Stripe
Designed for tissue culture work
Sterile, nonwettable pipets offer a tolerance of ±2% at full volume and have plug-in mouth end
  • Two pipet styles are available: one with magnifier stripe and one without
  • Each pipet lot is tested for sterility, accuracy and endotoxin level
CERTIFICATIONS
Certified nonpyrogenic with an endotoxin level less than 0.5EU/mL following USP XXII Bacterial Endotoxins Test (85).
COMPLIANCE
Calibrated “To Deliver/Blow Out”
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Sterile Polystyrene Disposable Serological Pipettes with Magnifier Stripe
  • 13-676-10M
  • 13-678-11 Series pipets have color-coded, transparent paper/plastic wrappers for easy identification and tear-through.
  • 13-676-10 and 13-678-11 Series
13-676-10M
(18 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
Capacity 2mL; Subdivision 1/100mL; Neg Grads 2/10mL; Tolerence ±0.04mL; Multipack; Green
Accessories
13-675-2C Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 500 for $163.30
Case of 500 for $152.52 Web Discount $10.78 (7%)
(0) in My Cart
Green; Capacity: 2mL
Accessories
13-675-3C Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 500 for $183.50
Case of 500 for $155.98 Web Discount $27.52 (15%)
(0) in My Cart
Yellow; 0.2mL negative graduations; Capacity: 1mL; 0.01mL Subdiv.
Accessories
13-676-10B Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 1000 for $257.00
(0) in My Cart
Subdivisions: 0.1mL; Multipack; Capacity: 5mL; 500/Cs.
Accessories
13-676-10C Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 500 for $196.70
Case of 500 for $177.23 Web Discount $19.47 (10%)
(0) in My Cart
Subdivisions: 0.1mL; Multipack; Capacity: 3mL; Orange; 500/Cs.
Accessories
13-676-10F Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 500 for $215.00
Case of 500 for $204.68 Web Discount $10.32 (5%)
(0) in My Cart
Yellow; Capacity: 1mL
Accessories
13-676-10G Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 1000 for $278.50
Case of 1000 for $250.65 Web Discount $27.85 (10%)
(0) in My Cart
Blue; Capacity: 5mL
Accessories
13-676-10H Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 200 for $91.50
Case of 200 for $86.38 Web Discount $5.12 (6%)
(0) in My Cart
All plastic; Orange; Capacity: 10mL; Individually packed
Accessories
13-676-10J Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 200 for $96.20
Case of 200 for $90.81 Web Discount $5.39 (6%)
(0) in My Cart
Capacity: 25mL
Accessories
13-676-10K Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 200 for $229.50
Case of 200 for $216.19 Web Discount $13.31 (6%)
(0) in My Cart
Subdivisions: 0.2mL; Multipack; Capacity: 25mL; Red; 200/Cs.
Accessories
13-676-10MAvailable on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 200 for $214.50
Case of 200 for $193.05 Web Discount $21.45 (10%)
(0) in My Cart
Capacity 50mL; Subdivision 5/10mL; Tolerence ±1mL; Multipack; Purple
Accessories
13-676-10Q Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 100 for $243.00
Case of 100 for $230.85 Web Discount $12.15 (5%)
(0) in My Cart
Purple; 50mL; ±1mL tolerance; 5/10mL Subdiv.
Accessories
13-676-10R Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 100 for $246.00
Case of 100 for $228.78 Web Discount $17.22 (7%)
(0) in My Cart
Red; 10mL negative graduations; Capacity: 25mL; 0.5mL subdiv.
Accessories
13-678-11 Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 200 for $202.00
Case of 200 for $181.80 Web Discount $20.20 (10%)
(0) in My Cart
Yellow; +/- 0.2 negative graduations; Capacity 1mL; 0.01mL subdiv.
Accessories
13-678-11B Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 1000 for $268.50
(0) in My Cart
Green; +/- 0.04 negative graduations; Capacity: 2mL; 0.01mL subdiv.
Accessories
13-678-11C Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 500 for $164.70
Case of 500 for $153.83 Web Discount $10.87 (7%)
(0) in My Cart
Blue; +/- 0.1 negative graduations; Capacity: 5mL; 0.1mL subdiv.
Accessories
13-678-11D Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 200 for $85.10
Case of 200 for $76.68 Web Discount $8.42 (10%)
(0) in My Cart
Orange; +/- 0.2 negative graduations; Capacity: 10mL; 0.1mL subdiv.
Accessories
13-678-11E Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 200 for $92.60
Case of 200 for $85.75 Web Discount $6.85 (7%)
(0) in My Cart
Subdivisions: 0.5mL; Individually packed; Capacity: 50mL; Purple; 100/Cs.
Accessories
13-678-11F Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 100 for $253.00
Case of 100 for $227.70 Web Discount $25.30 (10%)
(0) in My Cart
Add Items to Cart
Description
Sterile, nonwettable pipets offer a tolerance of ±2% at full volume and have plug-in mouth end
  • Two pipet styles are available: one with magnifier stripe and one without
  • Each pipet lot is tested for sterility, accuracy and endotoxin level
CERTIFICATIONS
Certified nonpyrogenic with an endotoxin level less than 0.5EU/mL following USP XXII Bacterial Endotoxins Test (85).
COMPLIANCE
Calibrated “To Deliver/Blow Out”
  • Black magnifier stripe runs lengthwise along pipet, making meniscus obvious at arm's length—reducing reading errors
  • Pipets have improved optical clarity—high-contrast black markings make volumes easier to read
  • Sterile and nonwettable pipets
  • Offer a tolerance of ±2% at full volume and have plug-in mouth end
  • 5, 10, 25, and 50mL pipets have ascending and descending graduations
  • Improved tip design on 25 and 50mL pipets increases the effect of surface tension to minimize dripping
  • Color coding on each pipet and on inner bags for multipack pipets make it easy to spot the size needed
  • Each pipet lot is tested for sterility, accuracy and endotoxin level
  • Individually wrapped, with paper and plastic wraps, 50 per pack/200 per case
Specifications & Ordering Information:
Inner bags are recyclable LDPE; outer boxes also are recyclable. Individually wrapped pipets are ideal for small-volume use requiring sterility and are available in either a clear, durable all-plastic wrap, or a color-coded paper/plastic wrap for reduced static cling. Multipack for larger-volume use permits easy access to several pipets at a time without contaminating unused pipets.
Certifications: Certified nonpyrogenic with an endotoxin level less than 0.5EU/mL following USP XXII Bacterial Endotoxins Test (85).
Compliance: Calibrated “To Deliver/Blow Out”
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.