Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
13-986-126A
View Image Gallery (4) Image(s) are representative of the product Group
  • 13-986-126A
  • 13-986-128S
  • 13-986-128S
  • Controller
Fisher Scientific™ Isotemp™ Plus Lab Refrigerators
Microprocessor-controlled temperatures in a choice of capacities and styles
  • Temperature range from 1° to 12°C (33.8° to 53.6°F) with ±3° average stability; adjustable in °C or °F using interior control
  • Exterior-mounted 0.5 in. LED display with 0.1° temperature resolution
  • Capacities from 23.1 to 74 cu. ft. (654 to 2100L)
  • Automatic defrost
Construction
  • Gray enamel-coated steel cabinets, stainless-steel interiors Keylocked solid or glass doors
  • Solid doors are hinged, white enamel-coated steel
  • Thermopane™ glass doors (hinged or sliding) provide easy access and visual inventory control
  • Sliding doors require no swing-out space—a valuable benefit in high traffic or narrow areas
  • Gray epoxy-coated steel-wire shelves (14.8 to 44.4 sq. ft.), adjustable on 1 in. (2.5cm) centers
  • CFC-free R134A refrigerant and polyurethane insulation
  • Interior fluorescent lighting with exterior light switch
  • Keyed ON/OFF power switch
  • Four adjustable legs ensure cabinets are level on irregular floors (casters provided)
Alarms
  • Panel-mounted, factory installed
  • Digital display with minimum/maximum setpoints, audible alarm, and 24V, 0.5A SPST N.O. relay contacts
  • Freestanding (13-986-160), Standard Remote (13-986-32A), and Deluxe Remote (13-986-100) alarm systems available separately
Seven-day Selectable Temperature Recorder
  • Panel-mounted, factory installed
  • 6 in. dia. (15.2cm) chart User-selectable temperature ranges
  • Single-point calibration; individually calibrated against equipment with calibration certified traceable to NIST™
  • Supplied with 60 charts and 7-day scale
View Specifications
Images for Fisher Scientific™ Isotemp™ Plus Lab Refrigerators
  • 13-986-126A
  • 13-986-128S
  • 13-986-128S
  • Controller
13-986-126A
13-986-126A
(2 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
13-986-132 Capacity: 74cu.ft.; Without recorder and alarm; Triple glass door
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
13-986-122 Capacity: 34cu.ft.; Without recorder and alarm; Solid door
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Description
  • Temperature range from 1° to 12°C (33.8° to 53.6°F) with ±3° average stability; adjustable in °C or °F using interior control
  • Exterior-mounted 0.5 in. LED display with 0.1° temperature resolution
  • Capacities from 23.1 to 74 cu. ft. (654 to 2100L)
  • Automatic defrost
Construction
  • Gray enamel-coated steel cabinets, stainless-steel interiors Keylocked solid or glass doors
  • Solid doors are hinged, white enamel-coated steel
  • Thermopane™ glass doors (hinged or sliding) provide easy access and visual inventory control
  • Sliding doors require no swing-out space—a valuable benefit in high traffic or narrow areas
  • Gray epoxy-coated steel-wire shelves (14.8 to 44.4 sq. ft.), adjustable on 1 in. (2.5cm) centers
  • CFC-free R134A refrigerant and polyurethane insulation
  • Interior fluorescent lighting with exterior light switch
  • Keyed ON/OFF power switch
  • Four adjustable legs ensure cabinets are level on irregular floors (casters provided)
Alarms
  • Panel-mounted, factory installed
  • Digital display with minimum/maximum setpoints, audible alarm, and 24V, 0.5A SPST N.O. relay contacts
  • Freestanding (13-986-160), Standard Remote (13-986-32A), and Deluxe Remote (13-986-100) alarm systems available separately
Seven-day Selectable Temperature Recorder
  • Panel-mounted, factory installed
  • 6 in. dia. (15.2cm) chart User-selectable temperature ranges
  • Single-point calibration; individually calibrated against equipment with calibration certified traceable to NIST™
  • Supplied with 60 charts and 7-day scale
Specifications & Ordering Information:
208/230V models equipped with NEMA L-1420P line-cord plug, require NEMA L14-20R receptacle. Voltages not for international use.
Certifications: UL listed
Warranty: 13 months, parts and labor
Service agreement available.
Specifications 
StyleUpright
Temperature Range1° to 12°C (33.8° to 53.6°F)
RefrigerantR-134a
Condenser TypeFin and Tube
DefrostAutomatic
CastersYes
InsulationPolyurethane foam
Plug TypeNEMA 5-15
Electrical Requirements115V/60Hz