Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Available on GSA/VA Contract for Federal Government customers only. Heidolph™ Tuttnauer™ Automated Electronic Benchtop Sterilizers
Compact sterilizers featuring electronic controls that provide fast, safe, and convenient way to sterilize both solids and liquids
Standard Operating Features
  • Self-contained design eliminates external plumbing and hardwiring for easy installation
  • Simple operation
  • ASME-stamped, Type 316 L stainless-steel chamber ensures long-lasting reliability, durability, and high-corrosion resistance
  • Fast/slow exhause (fast allows quick cycle time of solids while slow exhause prevents media and liquid boil-over)
  • Closed-door drying system maintains sterility
  • Drying cycle removes residual moisture after sterilization
  • Continuous pressure purge eliminates air pockets and ensures thorough, even sterilization
  • Large-capacity reservoir provides ample water supply to avoid frequent fill-ups
  • Water drain is conveniently located on top to allow quick and easy cleaning of reservoir
  • Automatic shut-off feature saves energy
Standard Safety Features
  • Heat-insulated, double-locking door protects against accidental burns
  • Door cannot be opened while chamber is under pressure
  • Automatic low water level shut-off prevents damage to the heater elements if there is insufficient water for the sterilization cycyle
  • Overtemperature cutoff and overpressure valve ensure safe operation
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Heidolph™ Tuttnauer™ Automated Electronic Benchtop Sterilizers
(22 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
02-964-41 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210665
3545E, 220v
Accessories
Each for $11,829.11
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
13-876-705 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210398
2540EAKP, 220V
Accessories
Each for $11,547.04
Check Availability
Add
to Cart
(0) in My Cart
13-889-473 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210258
2340E, 220V
Each for $7,237.07
Check Availability
Add
to Cart
(0) in My Cart
02-964-28 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210495
3850EP, 220v
Accessories
Each for $18,866.49
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
02-964-27 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210487
3850E, 220v
Accessories
Each for $18,528.24
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
13-889-474 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210282
2340EA, 115v
Each for $8,169.84
Check Availability
Add
to Cart
(0) in My Cart
13-876-704 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210380
2540EAP, 220V
Accessories
Each for $10,308.38
Check Availability
Add
to Cart
(0) in My Cart
02-964-11 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210452
2540E, 220v
Accessories
Each for $8,497.81
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
02-964-25 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210444
2540EKP, 220v
Accessories
Each for $10,122.33
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
02-964-42 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210690
3545EP, 220v
Accessories
Each for $12,522.81
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
02-964-14 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210568
3870EP, 220v
Accessories
Each for $24,436.64
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
02-964-5 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210207
2340E, 120V
Accessories
Each for $7,562.39
Check Availability
Add
to Cart
(0) in My Cart
13-889-475 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210291
2340EAP, 120V
Each for $8,667.23
Check Availability
Add
to Cart
(0) in My Cart
13-876-707 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210673
3870EAP, 220v
Accessories
Each for $24,209.15
Check Availability
Add
to Cart
(0) in My Cart
02-964-10B Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210606
3870E, 220v
Accessories
Each for $24,150.87
Check Availability
Add
to Cart
(0) in My Cart
13-876-702 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210330
2540EA, 115v
Accessories
Each for $9,241.05
Check Availability
Add
to Cart
(0) in My Cart
13-889-477 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210461
2540EP, 220V
Each for $8,638.86
Check Availability
Add
to Cart
(0) in My Cart
02-964-9 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210401
2540E, 120V
Accessories
Each for $8,746.29
Check Availability
Add
to Cart
(0) in My Cart
02 964-7 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210215
2340EP, 1400w, With Electronic Controls and Printer
Accessories
Each for $8,203.55
Check Availability
Add
to Cart
(0) in My Cart
02-964-10 Available on GSA/VA Contract for Federal Government customers only. 
Heidolph North America
No.:023210410
2540EP, 115v
Accessories
Each for $9,297.46
Check Availability
Add
to Cart
(0) in My Cart
13-876-701
Heidolph North America
No.:023210291X
2340EAP, 120V
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Part Not Found  Please contact customer service at 1-800-766-7000 with error code:999. 
13-876-700
Heidolph North America
No.:023210282X
2340EA, 120V
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Add Items to Cart
Description
Standard Operating Features
  • Self-contained design eliminates external plumbing and hardwiring for easy installation
  • Simple operation
  • ASME-stamped, Type 316 L stainless-steel chamber ensures long-lasting reliability, durability, and high-corrosion resistance
  • Fast/slow exhause (fast allows quick cycle time of solids while slow exhause prevents media and liquid boil-over)
  • Closed-door drying system maintains sterility
  • Drying cycle removes residual moisture after sterilization
  • Continuous pressure purge eliminates air pockets and ensures thorough, even sterilization
  • Large-capacity reservoir provides ample water supply to avoid frequent fill-ups
  • Water drain is conveniently located on top to allow quick and easy cleaning of reservoir
  • Automatic shut-off feature saves energy
Standard Safety Features
  • Heat-insulated, double-locking door protects against accidental burns
  • Door cannot be opened while chamber is under pressure
  • Automatic low water level shut-off prevents damage to the heater elements if there is insufficient water for the sterilization cycyle
  • Overtemperature cutoff and overpressure valve ensure safe operation
Applications: Sterilization of liquids, media, instruments, glassware, clothing, and waste
Automated Electronic Sterilizers
Electronic models use microprocessor controls to completely automate every stage of the cycle - water filling, heating, sterilization, exhaust and drying
Features of all automated electronic sterilizers
  • Sterilization time to 59 minutes
  • Temperature selection from 105° to 137°C (221° to 279°F)
  • In-line particle filter to protect valves
  • Electronic pressure and temperature sensors
  • Low-water alarm (sounds when reservoir needs to be refilled)
  • Standby heating keeps chamber warm for quicker cycles
EP Models
Helps meet GLP documentation requirements. Additional features include:
  • Built-in printer that provides a printed summary at the end of each cycle
  • Documents temperature, elapsed drying and sterilization time, selected cycle, cycle number, real time and date, errors or interruptions
  • Available with 9-, 10-, 12- and 15-inch diameter chambers
EA Models
Ideal for sterilizing wrapped and sealed instruments. Additional features include:
  • Fully automatic autoclaves are specially designed with preloaded cycles for sterilizing packs, pouches, sealed pipette racks, unwrapped instruments and other contained/wrapped materials
  • Highly efficient air-driven pump circulates hot air after the sterilization cycle to ensure thorough drying of samples and to reduce overall drying time
  • Available with 9-, 10- and 15-inch diameter chambers, with and without a built-in printer
Kwiklave Model 2540EAKP
  • Cuts sterilization time in half—perfect for fast instrument turnaround. Offers all standard operating and safety features, plus a special quick cycle
  • Advanced heating element reduces the time needed for the autoclave to reach optimal sterilization temperature, reducing overall sterilization time by up to 50%
  • Has a 10-inch diameter chamber and comes equipped with a built-in printer to document results
Certifications: UL listed, CSA, ASME (chamber)
Warranty: One year
Service agreement available.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.