Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
05-100-1
BD™ Clay Adams™ SeroFuge™ 2000 Series Centrifuges
Fixed- and dual-speed models for cell washing and serum separations
Ideal for blood grouping, typing, cross matching, and other cell washing procedures.
Includes: Unit and 12-place Plastic Rotor (05-100-7) for 12.7 dia. x 75mm blood tubes
  • Keypad controls allow precise speed and run time settings
  • Four-digit display for run time (up to 99 minutes, 99 seconds) and time remaining
  • Stored parameters allow repeatable runs
  • On-board tachometer
  • Supplied rotor has numbered slots for easy sample identification
  • Variable-control brake
  • Locking lid prevents lid opening until rotor has reached a complete stop
  • Gasketed lid seal protects operator from exposure to aerosols within rotor chamber
  • Durable molded polycarbonate base and lid with a white polyurethane enamel finish
  • Four cushioned, rubber feet minimize sound and vibration
View Specifications
Images for BD™ Clay Adams™ SeroFuge™ 2000 Series Centrifuges
  • 05-100-1
05-100-1
05-100-1
(3 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
05-100-1
BD Diagnostics
No.:420351
Model 2001; 12-place rotor; 115V
Each for $4,062.53
Check Availability
Add
to Cart
(0) in My Cart
05-100-3
BD Diagnostics
No.:420352
Model 2002; 12-place head rotor; 2-speed; 115V
Required Accessories
Each for $4,060.09
Check Availability
Add
to Cart
(0) in My Cart
05-100-2
BD Diagnostics
No.:420354
Model 2001; 12-place rotor; 1 speed; 230V
Required Accessories
N/A N/A N/A
This product is regulated by the federal government, and may not be ordered online. 
Add Items to Cart
Description
  • Keypad controls allow precise speed and run time settings
  • Four-digit display for run time (up to 99 minutes, 99 seconds) and time remaining
  • Stored parameters allow repeatable runs
  • On-board tachometer
  • Supplied rotor has numbered slots for easy sample identification
  • Variable-control brake
  • Locking lid prevents lid opening until rotor has reached a complete stop
  • Gasketed lid seal protects operator from exposure to aerosols within rotor chamber
  • Durable molded polycarbonate base and lid with a white polyurethane enamel finish
  • Four cushioned, rubber feet minimize sound and vibration
SeroFuge 2001 Fixed-speed Model
Max Speed: 3550rpm; Force: 1049xG
SeroFuge 2002 Dual-speed Model
Speed/force: 2400rpm/500xG or 3400rpm/1049xG
Specifications & Ordering Information:
Dimensions, all models: 17L x 13W x 12 in.H (43 x 33 x 30.5cm). Net weight: 28 lb. (12.7kg). For 115V 50/60Hz. Optional six-place aluminum rotor for plasma and serum separations (05-100-109) also available.
Service agreement available.