Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Powder-Free Latex Gloves
CTIS Correct-Touch™ Powder-Free Latex Gloves
No residual powder to contaminate environment or product
Natural rubber latex gloves are specially processed to control particles and contact transfer extractables to ensure ISO Class 5 (Class 100) compatibility. Processing also reduces proteins and chemicals known to cause skin irritation.
  • Ambidextrous
  • Double chlorinated to broaden chemical resistance
  • Textured surface provides a secure grip, wet or dry
  • Tapered, beaded cuff prevents rolldown and minimizes rips and tears during donning
View Specifications
Alert: These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Images for CTIS Correct-Touch™ Powder-Free Latex Gloves
  • Powder-Free Latex Gloves
Powder-Free Latex Gloves
Powder-Free Latex Gloves
(8 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
18-999-276A
CT International
No.:PFXT101
Length: 10 in. long (25cm); Size: Small
Pack of 100 for $38.75
Case of 10 PK for $348.90
Check Availability
Add
to Cart
(0) in My Cart
18-999-276B
CT International
No.:PFXT102
Length: 10 in. long (25cm); Size: Medium
Pack of 100 for $38.75
Case of 10 PK for $348.90
Check Availability
Add
to Cart
(0) in My Cart
18-999-276C
CT International
No.:PFXT103
Length: 10 in. long (25cm); Size: Large
Pack of 100 for $38.75
Case of 10 PK for $348.90
Check Availability
Add
to Cart
(0) in My Cart
18-999-276D
CT International
No.:PFXT104
Length: 10 in. long (25cm); Size: X-Large
Pack of 100 for $38.75
Case of 10 PK for $348.90
Check Availability
Add
to Cart
(0) in My Cart
18-999-277A
CT International
No.:PFXT201
Length: 12 in. long (30.5cm); Size: Small
Pack of 100 for $43.23
Case of 10 PK for $381.56
Check Availability
Add
to Cart
(0) in My Cart
18-999-277B
CT International
No.:PFXT202
Length: 12 in. long (30.5cm); Size: Medium
Pack of 100 for $43.23
Case of 10 PK for $381.56
Check Availability
Add
to Cart
(0) in My Cart
18-999-277C
CT International
No.:PFXT203
Length: 12 in. long (30.5cm); Size: Large
Pack of 100 for $43.23
Case of 10 PK for $381.56
Check Availability
Add
to Cart
(0) in My Cart
18-999-277D
CT International
No.:PFXT204
Length: 12 in. long (30.5cm); Size: X-Large
Pack of 100 for $43.23
Case of 10 PK for $381.56
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Ambidextrous
  • Double chlorinated to broaden chemical resistance
  • Textured surface provides a secure grip, wet or dry
  • Tapered, beaded cuff prevents rolldown and minimizes rips and tears during donning
Specifications & Ordering Information:
Leak tested. Double bagged.
Compliance: Meets or exceeds ASTM D 3578-95 standards. Sampled in accordance with MIL-STD-105E. Qualified for use in the pharmaceutical industry.