Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
19-149-863B
Get IT ItemKimberly-Clark Professional™ Purple Nitrile and Purple Nitrile-XTRA™ Exam Gloves
Unique purple color distinguishes these gloves from latex and other synthetic gloves
Contain no natural rubber latex proteins, low levels of chemical allergens.
  • Protect against a broad range of chemicals
  • Provide excellent tactile sensitivity
  • Textured fingertips enhance wet and dry grip
  • Powder- and odor-free
  • Single-use
  • Available in 9.5 in. and 12 in. lengths
  • Thickness: 5.9mil
  • Static dissipative in use
View Specifications
Images for Get IT ItemKimberly-Clark Professional™ Purple Nitrile and Purple Nitrile-XTRA™ Exam Gloves
  • 19-149-863B
19-149-863B
19-149-863B
(10 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
19-149-863 Get IT Item
Kimberly Clark
No.:55080
Length: 9.5 in.; X-Small
Pack of 100 for $32.60
Case of 10 PK for $265.80
Check Availability
Add
to Cart
(0) in My Cart
19-149-864 Get IT Item
Kimberly Clark
No.:55090
Length: 12 in.; X-Small
Pack of 50 for $36.36
Case of 10 PK for $306.41
Check Availability
Add
to Cart
(0) in My Cart
19-149-863A Get IT Item
Kimberly Clark
No.:55081
Length: 9.5 in.; Small
Pack of 100 for $32.22
Case of 10 PK for $262.91
Check Availability
Add
to Cart
(0) in My Cart
19-149-863B Get IT Item
Kimberly Clark
No.:55082
Length: 9.5 in.; Medium
Pack of 100 for $32.22
Case of 10 PK for $262.91
Check Availability
Add
to Cart
(0) in My Cart
19-149-863C Get IT Item
Kimberly Clark
No.:55083
Length: 9.5 in.; Large
Pack of 100 for $32.22
Case of 10 PK for $262.91
Check Availability
Add
to Cart
(0) in My Cart
19-149-863D Get IT Item
Kimberly Clark
No.:55084
Length: 9.5 in.; X-Large
Pack of 90 for $32.22
Case of 10 PK for $262.91
Check Availability
Add
to Cart
(0) in My Cart
19-149-864A Get IT Item
Kimberly Clark
No.:50601
Length: 12 in.; Small
Pack of 50 for $36.36
Case of 10 PK for $306.41
Check Availability
Add
to Cart
(0) in My Cart
19-149-864B Get IT Item
Kimberly Clark
No.:50602
Length: 12 in.; Medium
Pack of 50 for $36.36
Case of 10 PK for $306.41
Check Availability
Add
to Cart
(0) in My Cart
19-149-864C Get IT Item
Kimberly Clark
No.:50603
Length: 12 in.; Large
Pack of 50 for $36.36
Case of 10 PK for $306.41
Check Availability
Add
to Cart
(0) in My Cart
19-149-864D Get IT Item
Kimberly Clark
No.:50604
Length: 12 in.; X-Large
Pack of 50 for $36.36
Case of 10 PK for $306.41
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Protect against a broad range of chemicals
  • Provide excellent tactile sensitivity
  • Textured fingertips enhance wet and dry grip
  • Powder- and odor-free
  • Single-use
  • Available in 9.5 in. and 12 in. lengths
  • Thickness: 5.9mil
  • Static dissipative in use
Compliance: Exceed ASTM D 6319 standard. Meet AQL-1.5 for critical defects. Meet FDA requirement for Class I medical device.