Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
11-680-22
EchoTherm™ Benchtop Incubator, Chiller and Freezer
Five devices in one—water bath, dry bath, incubator, refrigerator, and freezer; Heats, chills, or freezes samples from -10° to +100°C—even at ambient temperatures; Energy-efficient Peltier design—no compressor or CFCs.
Includes: Includes RS-232-C interface for computer-directed data collection and unit control.A built-in datalogger can be programmed to collect temperature data points in 1-second, 1-minute, or 5-minute intervals to a total of 8110 data points. Includes a universal power supply that will accept inputs from 85 to 265 VAC, 50/60Hz and 12VDC output to chiller/heater module.
Compact, easy-to-use, and energy-efficient, Echotherm units are ideal for field or benchtop applications. Digital control from –10° to +100°C with ±1°C accuracy. Electronic calibration is factory set, but can be easily recalibrated to suit the user's needs. Two-line, alphanumeric, digital display is backlit for easy reading, shows target and actual temperatures continuously, and timer settings on demand. Integral countdown timer has a range from 1 second to 30 days, an audible alarm, and a user-settable auto-off feature. A buffer lamp on the front panel of the unit flashes when the datalogger buffer is collecting data and stays lit when the buffer is full of data to be downloaded. 50-watt power consumption.
View Specifications
Images for EchoTherm™ Benchtop Incubator, Chiller and Freezer
  • 11-680-22
11-680-22
11-680-22
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
11-680-22
Torrey Pines Scientific
No.:IC20
Incubator-Chiller-Freezer
Required Accessories
Each for $1,687.41
Check Availability
Add
to Cart
(0) in My Cart
Description
Compact, easy-to-use, and energy-efficient, Echotherm units are ideal for field or benchtop applications. Digital control from –10° to +100°C with ±1°C accuracy. Electronic calibration is factory set, but can be easily recalibrated to suit the user's needs. Two-line, alphanumeric, digital display is backlit for easy reading, shows target and actual temperatures continuously, and timer settings on demand. Integral countdown timer has a range from 1 second to 30 days, an audible alarm, and a user-settable auto-off feature. A buffer lamp on the front panel of the unit flashes when the datalogger buffer is collecting data and stays lit when the buffer is full of data to be downloaded. 50-watt power consumption.
  • RS-232-C interface for computer-directed data collection and unit control
  • Built-in datalogger collects temperature data points in 1-sec., 1-minute, or 5-minute intervals to a total of 8110 data points
Specifications & Ordering Information:
Small footprint, 83/4L x 61/2W x 31/2 in.H (22.2 x 16.5 x 8.9cm), takes up less bench space and is easily transported for remote applications. Aluminum sample blocks and clear polyethylene sample covers are available separately. Net weight (unit and power supply): 6.5 lb. (2.9kg). Aluminum chilling/heating surface measures 27/8 x 43/8 in. (7.3 x 11.1cm) and is very flat to facilitate uniform temperature transfer to samples.
Certifications: UL/CSA/CE approved.