Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
15-188-10
FisherBiotech™ MicroProbe™ Manual Staining System
Performs multistep staining protocols and simplifies standard chemical staining, immunostaining, and in situ hybridization
Includes: MicroProbe Staining Station with eight reagent buckets and lids; Locator tray for Isolon holders; Four Isolon holders; Five 30-well rubber reagent isolators; Five 3-well rubber reagent isolators; Six absorbent pads; slide holder with 72 ProbeOn microscope slides; MicroProbe Incubator with lid, calibration block, and power cord; User manual; Application guide
Staining Station
  • Eight reagent buckets with removable lids that can be filled to 20 or 100mL levels
  • Removable tray retains four plastic holders that accept Isolon™ reagent isolators or absorbent pads
Slide Holder Assembly and ProbeOn™ Microscope Slides
  • Allow users to move 20 ProbeOn slides (10 pairs) between staining station and incubator
  • Patented holder has lightweight, chemical-resistant plastic handle with numbered positions and indexing bump
View Specifications
Images for FisherBiotech™ MicroProbe™ Manual Staining System
  • 15-188-10
15-188-10
15-188-10
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
15-188-10 Configured for 120/240V, 50/60Hz
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Description
Staining Station
  • Eight reagent buckets with removable lids that can be filled to 20 or 100mL levels
  • Removable tray retains four plastic holders that accept Isolon™ reagent isolators or absorbent pads
Slide Holder Assembly and ProbeOn™ Microscope Slides
  • Allow users to move 20 ProbeOn slides (10 pairs) between staining station and incubator
  • Patented holder has lightweight, chemical-resistant plastic handle with numbered positions and indexing bump
Isolon™ Reagent Isolators
  • Available with three wells that contain milliliter volumes or with 30 smaller wells for microliter volumes of antibodies or DNA probes
  • Disposable, 30-well glass isolators are also available
MicroProbe Incubator
  • Heats from 30° to 110°C (86° to 230°F) in less than 10 minutes
  • Internal fan
  • Anodized aluminum incubation chamber
Compliance: CSA approved
Warranty: One year
† Customer configurable for 100/220/240VAC. (Line cords must be ordered separately—contact your Fisher Scientific Customer Service Representative.)