Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Fisher Scientific- null - 623148 - Product Details
10-650-14A
Fisher Scientific™ Isotemp™ Programmable Muffle Furnaces
Microprocessor control of linear heating and cooling rates and program duration; heat to 1125°C
Includes: 10-year lithium battery for memory backup
  • Available in three capacities
  • 24-step programmability plus one-program storage capability allows up to 12 heating/cooling ramps or dwell periods up to 99 hours 59 minutes each to be specified in any combination
  • LED displays furnace sequence, set temperature and actual temperature (°C or °F)
  • 24-ramp/soak profiles can be repeated continuously for an infinite number of cycles—furnace will automatically ensure that temperature at end of each segment is within a specified tolerance before the profile continues
  • One-program storage for frequently used run parameters
  • Auto shutoff with flashing alert at end of heating sequence
  • Continuous-run mode
  • Bottom and side-mounted heating elements are embedded in removable, molded alumina-silicate ceramic fiber panels for easy replacement
  • Light ceramic chamber makes unit easily portable; eliminates chipping and dust of traditional firebrick furnaces
  • Welded steel cabinet with baked-on finish
  • Solid hearth plate
Safety Features
  • Selectable restart after power interruption (Restart, Hold, or Shut Down modes)
  • Overtemperature protection with display code alerts if temperature deviates from 1°C to over 100°C from the setpoint (settable in 1°C increments), to a maximum of 1125°C (2057°F)
  • Asbestos-free multilayer, internal insulation keeps exterior surfaces cool to touch
  • Automatic heating shutoff when door is opened
  • Manually resettable circuit breaker protects internal components from power surges
  • Ventilated base prevents damage to benchtop
  • Top-mounted exhaust port allows easy venting
View Specifications
Images for Fisher Scientific™ Isotemp™ Programmable Muffle Furnaces
  • 10-650-14A
10-650-14A
10-650-14A
Description
  • Available in three capacities
  • 24-step programmability plus one-program storage capability allows up to 12 heating/cooling ramps or dwell periods up to 99 hours 59 minutes each to be specified in any combination
  • LED displays furnace sequence, set temperature and actual temperature (°C or °F)
  • 24-ramp/soak profiles can be repeated continuously for an infinite number of cycles—furnace will automatically ensure that temperature at end of each segment is within a specified tolerance before the profile continues
  • One-program storage for frequently used run parameters
  • Auto shutoff with flashing alert at end of heating sequence
  • Continuous-run mode
  • Bottom and side-mounted heating elements are embedded in removable, molded alumina-silicate ceramic fiber panels for easy replacement
  • Light ceramic chamber makes unit easily portable; eliminates chipping and dust of traditional firebrick furnaces
  • Welded steel cabinet with baked-on finish
  • Solid hearth plate
  • Safety Features
    • Selectable restart after power interruption (Restart, Hold, or Shut Down modes)
    • Overtemperature protection with display code alerts if temperature deviates from 1°C to over 100°C from the setpoint (settable in 1°C increments), to a maximum of 1125°C (2057°F)
    • Asbestos-free multilayer, internal insulation keeps exterior surfaces cool to touch
    • Automatic heating shutoff when door is opened
    • Manually resettable circuit breaker protects internal components from power surges
    • Ventilated base prevents damage to benchtop
    • Top-mounted exhaust port allows easy venting
    "/>
    Safety Features: Applications:
  • 0.14 cu. ft. (3.96L) model: 20 Fisherbrand™ quartz crucibles or up to 15 standard porcelain crucibles
  • 0.58 cu. ft. (16.4L) model: 84 Fisherbrand quartz crucibles or up to 50 standard porcelain crucibles (when equipped with optional 10-651SN Accessory Shelf)
  • 1.26 cu. ft. (35.7) model: 168 Fisherbrand quartz crucibles or up to 100 standard porcelain crucibles (when equipped with standard 10-750S Shelf and two 10-651SN Accessory Shelves)
  • 0.58 and 1.26 cu. ft. models have ceramic fiber rope door gaskets that prevent heat loss from the chamber
  • "/>
    Specifications & Ordering Information:
    Certifications:
    Compliance:
    Recommended Storage:
    Warranty:
    1 As performed per ASTM E 145.
    2 At 500° to 1125°C (932° to 2057°F).
    3 Accuracy offset differences are customer adjustable by ±30°C (±54°F).
    4 For loads over 3 lb. (1.4kg), replace 10-750S shelf with 10-650H solid hearth plate. Contact your Fisher Scientific Representative for details.
    5 CSA approved.
    6 This unit is suitable as an International Model.