Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Left to right: 11-394-305, 11-394-200, 19-059-925
View Image Gallery (1) Image(s) are representative of the product Group
  • Left to right: 11-394-305, 11-394-200, 19-059-925
Tempshield* Cryo-Gloves*
Breathable, stay-dry lining
Ideal for use in cryogenic atmospheres (such as liquid nitrogen, oxygen or helium; liquified natural gas; dry ice; liquid baths; or blast freezers) where organs, blood units, pharmaceuticals, or specimens are frozen.
  • Comfortable and lightweight with excellent dexterity
  • Protect hands and arms in temperatures as low as -162°C (-260°F)
  • Water-resistant and waterproof styles with breathable, stay-dry lining
  • Contain no asbestos
View Specifications
Alert: Not for immersion in liquid nitrogen or for use near open flame or in hazardous areas where ignition could occur.
Images for Tempshield* Cryo-Gloves*
  • Left to right: 11-394-305, 11-394-200, 19-059-925
Left to right: 11-394-305, 11-394-200, 19-059-925
(23 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
Water-resistant midarm length with liner; Size: Large
11-394-200
Tempshield
No.:MAL
N/AYou must be logged in and have a valid account number to view item availability   Pair for $168.90
Case of 10 PR for $1,344.00
(0) in My Cart
Water-resistant midarm length with liner; Size: Medium
11-394-305
Tempshield
No.:MAM
N/AYou must be logged in and have a valid account number to view item availability   Pair for $168.90
Case of 10 PR for $1,344.00
(0) in My Cart
Water-resistant elbow length with liner; Size: Medium
11-394-306
Tempshield
No.:EBM
N/AYou must be logged in and have a valid account number to view item availability   Pair for $210.50
Case of 10 PR for $1,776.00
(0) in My Cart
Water-resistant elbow length with liner; Size: Large
11-394-307
Tempshield
No.:EBL
N/AYou must be logged in and have a valid account number to view item availability   Pair for $210.50
Case of 10 PR for $1,674.00
(0) in My Cart
Water-resistant shoulder length with liner; Size: Large
11-394-308
Tempshield
No.:SHL
N/AYou must be logged in and have a valid account number to view item availability   Pair for $279.00
Case of 10 PR for $2,230.00
(0) in My Cart
Water-resistant elbow length with liner; Size: XL
11-394-309
Tempshield
No.:EBXL
N/AYou must be logged in and have a valid account number to view item availability   Pair for $210.50
Case of 10 PR for $1,720.00
(0) in My Cart
Waterproof wrist length with liner; Size: Small
11-394-322
Tempshield
No.:WRSWP
N/AYou must be logged in and have a valid account number to view item availability   Pair for $193.30
Case of 10 PR for $1,546.00
(0) in My Cart
Waterproof wrist length with liner; Size: Medium
11-394-323
Tempshield
No.:WRMWP
N/AYou must be logged in and have a valid account number to view item availability   Pair for $193.30
Case of 10 PR for $1,544.00
(0) in My Cart
Waterproof wrist length with liner; Size: Large
11-394-324
Tempshield
No.:WRLWP
N/AYou must be logged in and have a valid account number to view item availability   Pair for $193.30
Case of 10 PR for $1,544.00
(0) in My Cart
Waterproof midarm length with liner; Size: Small
11-394-325
Tempshield
No.:MASWP
N/AYou must be logged in and have a valid account number to view item availability   Pair for $208.50
Case of 10 PR for $1,664.00
(0) in My Cart
Waterproof midarm length with liner; Size: Medium
11-394-326
Tempshield
No.:MAMWP
N/AYou must be logged in and have a valid account number to view item availability   Pair for $208.50
Case of 10 PR for $1,657.00
(0) in My Cart
Waterproof midarm length with liner; Size: Large
11-394-327
Tempshield
No.:MALWP
N/AYou must be logged in and have a valid account number to view item availability   Pair for $208.50
Case of 10 PR for $1,650.00
(0) in My Cart
Waterproof midarm length with liner; Size: XL
11-394-328
Tempshield
No.:MAXLWP
N/AYou must be logged in and have a valid account number to view item availability   Pair for $208.50
Case of 10 PR for $1,649.00
(0) in My Cart
Waterproof elbow length with liner; Size: Medium
11-394-329
Tempshield
No.:EBMWP
N/AYou must be logged in and have a valid account number to view item availability   Pair for $257.50
Pair for $244.63 Web Discount $12.87 (5%)
Case of 10 PR for $2,050.00
Case of 10 PR for $1,947.50 Web Discount $102.50 (5%)
(0) in My Cart
Waterproof elbow length with liner; Size: Large
11-394-330
Tempshield
No.:EBLWP
N/AYou must be logged in and have a valid account number to view item availability   Pair for $257.50
Case of 10 PR for $2,050.00
(0) in My Cart
Waterproof elbow length with liner; Size: XL
11-394-331
Tempshield
No.:EBXLWP
N/AYou must be logged in and have a valid account number to view item availability   Pair for $257.50
Case of 10 PR for $2,060.00
(0) in My Cart
Water-resistant midarm length with liner; Size: XL
19-059-925
Tempshield
No.:MAXL
N/AYou must be logged in and have a valid account number to view item availability   Pair for $225.50
Pair for $214.23 Web Discount $11.27 (5%)
(0) in My Cart
To shoulder; X- Large
19-159-178
TEMPSHIELD INC
No.:SHXL
N/AYou must be logged in and have a valid account number to view item availability   Pair for $372.00
(0) in My Cart
Water-resistant wrist length with liner; Size: Small
19-276-533
Tempshield
No.:WRS
N/AYou must be logged in and have a valid account number to view item availability   Pair for $157.70
Pair for $149.82 Web Discount $7.88 (5%)
Case of 10 PR for $1,348.00
Case of 10 PR for $1,280.60 Web Discount $67.40 (5%)
(0) in My Cart
Water-resistant wrist length with liner; Size: Medium
19-276-534
Tempshield
No.:WRM
N/AYou must be logged in and have a valid account number to view item availability   Pair for $157.70
Case of 10 PR for $1,260.00
(0) in My Cart
Water-resistant wrist length with liner; Size: Large
19-276-535
Tempshield
No.:WRL
N/AYou must be logged in and have a valid account number to view item availability   Pair for $157.70
Case of 10 PR for $1,260.00
(0) in My Cart
To shoulder; Large
19-276-547
TEMPSHIELD INC
No.:SHLWP
N/AYou must be logged in and have a valid account number to view item availability   Pack for $438.00
Pack for $416.10 Web Discount $21.90 (5%)
(0) in My Cart
Small; To elbow
11-394-306A
TEMPSHIELD INC
No.:EBS
N/AYou must be logged in and have a valid account number to view item availability   Case of 10 for $2,110.00
(0) in My Cart
Add Items to Cart
Description
  • Comfortable and lightweight with excellent dexterity
  • Protect hands and arms in temperatures as low as -162°C (-260°F)
  • Water-resistant and waterproof styles with breathable, stay-dry lining
  • Contain no asbestos