Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
13-986-149
View Image Gallery (1) Image(s) are representative of the product Group
  • 13-986-149
Available on GSA/VA Contract for Federal Government customers only. Fisher Scientific* Isotemp* Flammable-Materials Storage Freezers
For storing volatile materials, pharmaceuticals, clinical reagents, and biological samples
20.6 cu. ft. (583L) Model
  • Temperature range: -20° to -12°C (average stability ±3°C)
  • Manual defrost
  • Four fixed shelves (13.5 sq. ft. capacity)
Safety Features
  • Sparkproof interiors separate contents from compressor and motor
  • Top-mounted hydraulic thermostat and door stop
  • Magnetic door gaskets provide airtight seals that insulate and prevent release of hazardous fumes
  • Compressors have enclosed overloads, relays, and thermostats to prevent sparks from escaping during opening and closing
  • Static hot-wall condensers and evaporators are free of electric fans
Construction
  • One hinged solid door with key lock
  • White, baked-on polyester enamel exterior with orange warning label
  • White epoxy-coated steel interior with ABS plastic door liner
  • CFC-free R404A refrigerant, polyurethane foam insulation
View Specifications
Alert: Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Images for Available on GSA/VA Contract for Federal Government customers only. Fisher Scientific* Isotemp* Flammable-Materials Storage Freezers
  • 13-986-149
13-986-149
(1 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
Basic; 20.6 cu. ft.; -20 to -12°C
MSDS
Accessories
13-986-149 Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Each for $3,660.00
(0) in My Cart
Description
20.6 cu. ft. (583L) Model
  • Temperature range: -20° to -12°C (average stability ±3°C)
  • Manual defrost
  • Four fixed shelves (13.5 sq. ft. capacity)
Safety Features
  • Sparkproof interiors separate contents from compressor and motor
  • Top-mounted hydraulic thermostat and door stop
  • Magnetic door gaskets provide airtight seals that insulate and prevent release of hazardous fumes
  • Compressors have enclosed overloads, relays, and thermostats to prevent sparks from escaping during opening and closing
  • Static hot-wall condensers and evaporators are free of electric fans
Construction
  • One hinged solid door with key lock
  • White, baked-on polyester enamel exterior with orange warning label
  • White epoxy-coated steel interior with ABS plastic door liner
  • CFC-free R404A refrigerant, polyurethane foam insulation
Specifications & Ordering Information:
For 115V 60Hz. Supplied with NEMA 5-15P power cord; requires NEMA 5-15R wall receptacle. Freestanding (13-986-160), Standard Remote (13-986-32A), and Deluxe Remote (13-986-100) alarm systems available separately.
Certifications: UL listed, CSA approved
Warranty: 13 months, parts and labor
Service agreement available.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.