Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
1450910
View Image Gallery (2) Image(s) are representative of the product Group
  • 1450910
  • 1450911
Waring™ Laboratory Blenders: Single Speed
Meet the requirements of the most demanding mixing and blending applications
Includes: Base, container, 3-wire grounded cord and plug
  • Single-speed
  • Cloverleaf design
  • Choice of glass and/or stainless-steel containers
  • Sturdy, diecast metal bases and stainless-steel blades
  • All durably constructed for years of dependable service
  • Epoxy-coated bases with chrome top pieces on all models, except for the aluminum and steel base (Hazardous-Location Model)
  • Heat-resistant borosilicate glass (integral handle) or stainless-steel (attached handles)
  • Two-piece vinyl and styrene lids
View Specifications
Images for Waring™ Laboratory Blenders: Single Speed
  • 1450910
  • 1450911
1450910
1450910
(3 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
14-509-10
Conair Corporation
No.:700G
One-speed; 1L (34 oz.) Glass container; Single speed commercial; 120V
Accessories
N/A N/A N/A
Price File Not Open 
14-509-50
Conair Corporation
No.:700S
One-speed; 1L (34 oz.) Stainless-steel container; Commercial; 120V
Accessories
N/A N/A N/A
Price File Not Open 
14-509-11
Conair Corporation
No.:800G
Single-speed; 1L (34 oz.) Glass container; Heavy Duty; 240V
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Price File Not Open 
Description
  • Single-speed
  • Cloverleaf design
  • Choice of glass and/or stainless-steel containers
  • Sturdy, diecast metal bases and stainless-steel blades
  • All durably constructed for years of dependable service
  • Epoxy-coated bases with chrome top pieces on all models, except for the aluminum and steel base (Hazardous-Location Model)
  • Heat-resistant borosilicate glass (integral handle) or stainless-steel (attached handles)
  • Two-piece vinyl and styrene lids
Specifications & Ordering Information:
Electrical requirements for all models: 115/120V 50/60Hz
Certifications: UL listed (Select models only; see ordering chart)